Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03649943_m1
          See other BC005561 GE Assays ›
          Gene Symbol:
          BC005561
          Gene Name:
          cDNA sequence BC005561
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 104508352 - 104522383 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03649943_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC005561
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001166581.1 NP_001160053.1 1 - 2 222 127
          GenBank mRNA AK164034.1 - 1 - 2 223 127
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CO043956.1 - 1 - 2 Chr. 5 - 104508415 - 104517813 [-] on Build GRCm38 127
          GenBankmRNA BY729033.1 - 1 - 2 223 Chr. 5 - 104508352 - 104517875 [+] on Build GRCm38 127
          GenBankmRNA BU840354.1 - 195 Chr. - - [-] on Build 127

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream polycystic kidney disease 2 Pkd2 C030034P18Rik,
          PC2,
          TRPP2
          -2.5kb Sense Chr. 5 - 104457224 - 104505819 on NCBI Build GRCm38
          Downstream RIKEN cDNA D930016D06 gene D930016D06Rik 100041172,
          1110004B06Rik,
          AI451450,
          Gm3179
          3.4kb Sense Chr. 5 - 104525735 - 104554211 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001028.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK164034.1, NM_001166581.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2360427 AK134379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2360427 NM_001166581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2360427 AK134379.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2360427 NM_001166581.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04920139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23952698 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1427461_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523746 - -
          Mouse Genome 430 2.0 Array 1427461_at - -
          Mouse Genome 430 2.0 Array 1427461_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042165_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042165_at - -
          Affymetrix Mouse Clariom S Array PSR0500008691.mm.1 TC0500001028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008691.mm.1 NM_001166581.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008691.mm.1 NM_001166581.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008692.mm.1 TC0500001028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500008692.mm.1 NM_001166581.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500008692.mm.1 NM_001166581.1 Refseq
          Mouse Expression Set 430 1427461_PM_at - -
          Mouse Expression Set 430 1427461_at - -
          Mouse Genome 430A 2.0 Array 1427461_PM_at - -
          Mouse Genome 430A 2.0 Array 1427461_at - -
          Affymetrix HT Mouse Genome 430A Array 1427461_at - -
          Affymetrix HT Mouse Genome 430A Array 1427461_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix