Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03652156_s1
          See other RPS4L GE Assays ›
          Gene Symbol:
          Rps4l
          Gene Name:
          ribosomal protein S4-like
          Gene Aliases:
          1110033J19Rik, AA407494, Rps4y2
          Chromosome Location:
          Chr.6: 148354656 - 148355596 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03652156_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps4l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003634.2 1 149 104
          GenBank mRNA AK004068.1 - 1 150 104
          AK132160.1 - 1 150 104
          BC047994.1 - 1 121 104
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV511384.1 - 1 133 Chr. 6 - 148354670 - 148355107 [+] on Build GRCm38 104
          GenBankmRNA AV575050.1 - 1 85 Chr. 6 - 148354721 - 148355029 [+] on Build GRCm38 104
          GenBankmRNA AV578314.1 - 1 103 Chr. 6 - 148354702 - 148355170 [+] on Build GRCm38 104
          GenBankmRNA CK627377.1 - 1 76 Chr. 6 - 148354729 - 148355367 [+] on Build GRCm38 104
          GenBankmRNA CB204246.1 - 1 118 Chr. 6 - 148354687 - 148355578 [+] on Build GRCm38 104
          GenBankmRNA AV463957.1 - 1 121 Chr. 6 - 148354683 - 148355197 [+] on Build GRCm38 104
          GenBankmRNA AV516577.1 - 1 103 Chr. 6 - 148354706 - 148355172 [+] on Build GRCm38 104
          GenBankmRNA AV471013.1 - 1 126 Chr. 6 - 148354679 - 148355177 [+] on Build GRCm38 104
          GenBankmRNA AV507738.1 - 1 139 Chr. 6 - 148354667 - 148355175 [+] on Build GRCm38 104
          GenBankmRNA BY328788.1 - 1 Chr. 6 - 148354658 - 148354990 [+] on Build GRCm38 104
          GenBankmRNA AV448401.1 - 1 79 Chr. 6 - 148354726 - 148355221 [+] on Build GRCm38 104
          GenBankmRNA AV514849.1 - 1 139 Chr. 6 - 148354667 - 148355173 [+] on Build GRCm38 104
          GenBankmRNA CJ163072.1 - 1 147 Chr. 6 - 148354658 - 148355047 [+] on Build GRCm38 104
          GenBankmRNA CK022865.1 - 154 Chr. - - [-] on Build 104
          GenBankmRNA AV462502.1 - 1 129 Chr. 6 - 148354677 - 148355103 [+] on Build GRCm38 104
          GenBankmRNA CK137553.1 - 1 136 Chr. 6 - 148354669 - 148355260 [+] on Build GRCm38 104
          GenBankmRNA BG100415.1 - 1 67 Chr. 6 - 148354738 - 148355409 [+] on Build GRCm38 104
          GenBankmRNA AV467689.1 - 1 101 Chr. 6 - 148354709 - 148355103 [+] on Build GRCm38 104
          GenBankmRNA BX516077.1 - 1 134 Chr. 6 - 148354672 - 148355178 [+] on Build GRCm38 104
          GenBankmRNA AV511272.1 - 1 137 Chr. 6 - 148354667 - 148355095 [+] on Build GRCm38 104
          GenBankmRNA AV577061.1 - 1 136 Chr. 6 - 148354669 - 148355212 [+] on Build GRCm38 104
          GenBankmRNA AV475843.1 - 1 135 Chr. 6 - 148354668 - 148354978 [+] on Build GRCm38 104
          GenBankmRNA BY114163.1 - 1 147 Chr. 6 - 148354658 - 148354973 [+] on Build GRCm38 104
          GenBankmRNA AV472928.1 - 1 104 Chr. 6 - 148354701 - 148355113 [+] on Build GRCm38 104
          GenBankmRNA AV469257.1 - 1 91 Chr. 6 - 148354714 - 148355201 [+] on Build GRCm38 104
          GenBankmRNA AV463810.1 - 1 139 Chr. 6 - 148354667 - 148355103 [+] on Build GRCm38 104
          GenBankmRNA AV569749.1 - 1 102 Chr. 6 - 148354709 - 148355277 [+] on Build GRCm38 104
          GenBankmRNA AA007834.1 - 1 134 Chr. 6 - 148354671 - 148355250 [+] on Build GRCm38 104
          GenBankmRNA BY115419.1 - 1 147 Chr. 6 - 148354658 - 148354996 [+] on Build GRCm38 104
          GenBankmRNA AA041632.1 - 1 98 Chr. 6 - 148354707 - 148355194 [+] on Build GRCm38 104
          GenBankmRNA AV516234.1 - 1 118 Chr. 6 - 148354691 - 148355068 [+] on Build GRCm38 104
          GenBankmRNA AV493656.1 - 1 134 Chr. 6 - 148354670 - 148355111 [+] on Build GRCm38 104
          GenBankmRNA CB182173.1 - 1 145 Chr. 6 - 148354686 - 148355435 [+] on Build GRCm38 104
          GenBankmRNA AV483695.1 - 1 128 Chr. 6 - 148354682 - 148355097 [+] on Build GRCm38 104
          GenBankmRNA AV569452.1 - 1 125 Chr. 6 - 148354690 - 148355227 [+] on Build GRCm38 104
          GenBankmRNA AV453406.1 - 1 136 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38 104
          GenBankmRNA AV446887.1 - 1 139 Chr. 6 - 148354667 - 148355149 [+] on Build GRCm38 104
          GenBankmRNA BU841056.1 - 1 120 Chr. 6 - 148354685 - 148355589 [+] on Build GRCm38 104
          GenBankmRNA AV472654.1 - 1 124 Chr. 6 - 148354681 - 148355143 [+] on Build GRCm38 104
          GenBankmRNA AV583923.1 - 1 79 Chr. 6 - 148354726 - 148355242 [+] on Build GRCm38 104
          GenBankmRNA AV508759.1 - 1 130 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38 104
          GenBankmRNA AV587336.1 - 1 132 Chr. 6 - 148354672 - 148355188 [+] on Build GRCm38 104
          GenBankmRNA AV583728.1 - 1 130 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38 104
          GenBankmRNA BI525490.1 - 1 103 Chr. 6 - 148354709 - 148355592 [+] on Build GRCm38 104
          GenBankmRNA CJ171908.1 - 1 119 Chr. 6 - 148354687 - 148355065 [+] on Build GRCm38 104
          GenBankmRNA BU562610.1 - 1 133 Chr. 6 - 148354672 - 148355453 [+] on Build GRCm38 104
          GenBankmRNA AV515777.1 - 1 139 Chr. 6 - 148354667 - 148355103 [+] on Build GRCm38 104
          GenBankmRNA AV496116.1 - 1 133 Chr. 6 - 148354672 - 148355008 [+] on Build GRCm38 104
          GenBankmRNA AV506963.1 - 1 130 Chr. 6 - 148354677 - 148355017 [+] on Build GRCm38 104
          GenBankmRNA AA269378.1 - 1 64 Chr. 6 - 148354741 - 148355244 [+] on Build GRCm38 104
          GenBankmRNA AV475819.1 - 1 140 Chr. 6 - 148354673 - 148355149 [+] on Build GRCm38 104
          GenBankmRNA CB840729.1 - 1 130 Chr. 6 - 148354685 - 148355342 [+] on Build GRCm38 104
          GenBankmRNA AV452155.1 - 1 100 Chr. 6 - 148354705 - 148355157 [+] on Build GRCm38 104
          GenBankmRNA CK020541.1 - 1 121 Chr. 6 - 148354684 - 148355385 [+] on Build GRCm38 104
          GenBankmRNA AV506996.1 - 1 132 Chr. 6 - 148354682 - 148355099 [+] on Build GRCm38 104
          GenBankmRNA AV481267.1 - 1 134 Chr. 6 - 148354670 - 148355172 [+] on Build GRCm38 104
          GenBankmRNA AV467263.1 - 1 120 Chr. 6 - 148354684 - 148355103 [+] on Build GRCm38 104
          GenBankmRNA AV472068.1 - 1 131 Chr. 6 - 148354680 - 148355149 [+] on Build GRCm38 104
          GenBankmRNA BU551583.1 - 1 97 Chr. 6 - 148354708 - 148355277 [+] on Build GRCm38 104
          GenBankmRNA AV572982.1 - 1 132 Chr. 6 - 148354674 - 148355113 [+] on Build GRCm38 104
          GenBankmRNA CJ041563.1 - 1 150 Chr. 6 - 148354660 - 148355242 [+] on Build GRCm38 104
          GenBankmRNA AV499950.1 - 1 125 Chr. 6 - 148354679 - 148355085 [+] on Build GRCm38 104
          GenBankmRNA CO797874.1 - 111 Chr. - - [-] on Build 104
          GenBankmRNA AV494405.1 - 1 40 Chr. 6 - 148354765 - 148354985 [+] on Build GRCm38 104
          GenBankmRNA BY321323.1 - 1 95 Chr. 6 - 148354710 - 148355056 [+] on Build GRCm38 104
          GenBankmRNA AV472933.1 - 1 126 Chr. 6 - 148354685 - 148355069 [+] on Build GRCm38 104
          GenBankmRNA AV443982.1 - 1 126 Chr. 6 - 148354682 - 148355170 [+] on Build GRCm38 104
          GenBankmRNA AV587493.1 - 1 91 Chr. 6 - 148354723 - 148355138 [+] on Build GRCm38 104
          GenBankmRNA AI325738.1 - 1 84 Chr. 6 - 148354721 - 148355098 [+] on Build GRCm38 104
          GenBankmRNA AV475406.1 - 1 105 Chr. 6 - 148354700 - 148355189 [+] on Build GRCm38 104
          GenBankmRNA BU898844.1 - 1 133 Chr. 6 - 148354672 - 148355253 [+] on Build GRCm38 104
          GenBankmRNA AV494619.1 - 1 137 Chr. 6 - 148354668 - 148355181 [+] on Build GRCm38 104
          GenBankmRNA CK031884.1 - 1 113 Chr. 6 - 148354692 - 148355436 [+] on Build GRCm38 104
          GenBankmRNA AW701596.1 - 1 71 Chr. 6 - 148354734 - 148355262 [+] on Build GRCm38 104
          GenBankmRNA AV466506.1 - 1 99 Chr. 6 - 148354706 - 148355142 [+] on Build GRCm38 104
          GenBankmRNA CA321842.1 - 1 98 Chr. 6 - 148354706 - 148355409 [+] on Build GRCm38 104
          GenBankmRNA AA000082.1 - 1 143 Chr. 6 - 148354672 - 148355194 [+] on Build GRCm38 104
          GenBankmRNA BF153001.1 - 1 155 Chr. 6 - 148354672 - 148355081 [+] on Build GRCm38 104
          GenBankmRNA AV510526.1 - 1 135 Chr. 6 - 148354667 - 148355092 [+] on Build GRCm38 104
          GenBankmRNA AV498101.1 - 1 142 Chr. 6 - 148354662 - 148355099 [+] on Build GRCm38 104
          GenBankmRNA AV584069.1 - 1 132 Chr. 6 - 148354677 - 148355061 [+] on Build GRCm38 104
          GenBankmRNA AV482075.1 - 1 124 Chr. 6 - 148354682 - 148355090 [+] on Build GRCm38 104
          GenBankmRNA AV511109.1 - 1 137 Chr. 6 - 148354668 - 148355135 [+] on Build GRCm38 104
          GenBankmRNA AV494722.1 - 1 136 Chr. 6 - 148354668 - 148355191 [+] on Build GRCm38 104
          GenBankmRNA AV474962.1 - 1 132 Chr. 6 - 148354682 - 148355157 [+] on Build GRCm38 104
          GenBankmRNA AV463010.1 - 1 129 Chr. 6 - 148354674 - 148355148 [+] on Build GRCm38 104
          GenBankmRNA CJ164673.1 - 1 147 Chr. 6 - 148354658 - 148355078 [+] on Build GRCm38 104
          GenBankmRNA CB274693.1 - 2 100 Chr. 6 - 148354671 - 148355123 [+] on Build GRCm38 104
          GenBankmRNA AV451715.1 - 1 Chr. 6 - 148354669 - 148355098 [+] on Build GRCm38 105
          GenBankmRNA AV513781.1 - 1 90 Chr. 6 - 148354713 - 148355151 [+] on Build GRCm38 104
          GenBankmRNA AA028728.1 - 70 Chr. - - [-] on Build 104
          GenBankmRNA AV459279.1 - 1 135 Chr. 6 - 148354670 - 148355154 [+] on Build GRCm38 104
          GenBankmRNA AV581989.1 - 1 127 Chr. 6 - 148354678 - 148354982 [+] on Build GRCm38 104
          GenBankmRNA AV481595.1 - 1 128 Chr. 6 - 148354677 - 148355195 [+] on Build GRCm38 104
          GenBankmRNA AV464639.1 - 1 121 Chr. 6 - 148354683 - 148355133 [+] on Build GRCm38 104
          GenBankmRNA CK021457.1 - 1 116 Chr. 6 - 148354688 - 148355532 [+] on Build GRCm38 104
          GenBankmRNA AV484927.1 - 1 125 Chr. 6 - 148354678 - 148355189 [+] on Build GRCm38 104
          GenBankmRNA CJ040911.1 - 1 148 Chr. 6 - 148354658 - 148355273 [+] on Build GRCm38 104
          GenBankmRNA AV573175.1 - 1 127 Chr. 6 - 148354685 - 148355149 [+] on Build GRCm38 104
          GenBankmRNA AL117823.1 - 1 143 Chr. 6 - 148354667 - 148354928 [+] on Build GRCm38 104
          GenBankmRNA AV493615.1 - 1 95 Chr. 6 - 148354712 - 148355196 [+] on Build GRCm38 104
          GenBankmRNA BF012952.1 - 1 70 Chr. 6 - 148354735 - 148355078 [+] on Build GRCm38 104
          GenBankmRNA AV486209.1 - 1 140 Chr. 6 - 148354667 - 148355154 [+] on Build GRCm38 104
          GenBankmRNA AV488831.1 - 1 105 Chr. 6 - 148354705 - 148355197 [+] on Build GRCm38 104
          GenBankmRNA AV451783.1 - 1 130 Chr. 6 - 148354674 - 148355143 [+] on Build GRCm38 104
          GenBankmRNA BY096065.1 - 1 126 Chr. 6 - 148354680 - 148355071 [+] on Build GRCm38 104
          GenBankmRNA AV493432.1 - 1 107 Chr. 6 - 148354698 - 148355087 [+] on Build GRCm38 104
          GenBankmRNA CJ041148.1 - 1 105 Chr. 6 - 148354703 - 148355150 [+] on Build GRCm38 104
          GenBankmRNA AV573487.1 - 1 121 Chr. 6 - 148354684 - 148355133 [+] on Build GRCm38 104
          GenBankmRNA AV586344.1 - 1 95 Chr. 6 - 148354709 - 148355136 [+] on Build GRCm38 104
          GenBankmRNA AV480251.1 - 1 135 Chr. 6 - 148354685 - 148355145 [+] on Build GRCm38 104
          GenBankmRNA AV461169.1 - 1 128 Chr. 6 - 148354685 - 148355133 [+] on Build GRCm38 104
          GenBankmRNA C89050.1 - 150 Chr. - - [-] on Build 104
          GenBankmRNA AV465962.1 - 1 137 Chr. 6 - 148354667 - 148354985 [+] on Build GRCm38 104
          GenBankmRNA AV506464.1 - 1 130 Chr. 6 - 148354685 - 148355100 [+] on Build GRCm38 104
          GenBankmRNA AV455874.1 - 1 137 Chr. 6 - 148354668 - 148355142 [+] on Build GRCm38 104
          GenBankmRNA AV585951.1 - 1 69 Chr. 6 - 148354735 - 148355200 [+] on Build GRCm38 104
          GenBankmRNA AV503345.1 - 1 126 Chr. 6 - 148354682 - 148355096 [+] on Build GRCm38 104
          GenBankmRNA AV495870.1 - 1 134 Chr. 6 - 148354672 - 148355169 [+] on Build GRCm38 104
          GenBankmRNA DV662648.1 - 1 138 Chr. 6 - 148354667 - 148355422 [+] on Build GRCm38 104
          GenBankmRNA CB183113.1 - 1 126 Chr. 6 - 148354679 - 148355475 [+] on Build GRCm38 104
          GenBankmRNA CB845206.2 - 1 148 Chr. 6 - 148354667 - 148355453 [+] on Build GRCm38 104
          GenBankmRNA CK031938.1 - 1 113 Chr. 6 - 148354692 - 148355385 [+] on Build GRCm38 104
          GenBankmRNA AV464024.1 - 1 119 Chr. 6 - 148354683 - 148355097 [+] on Build GRCm38 104
          GenBankmRNA AV491120.1 - 1 128 Chr. 6 - 148354677 - 148355193 [+] on Build GRCm38 104
          GenBankmRNA AV485052.1 - 1 121 Chr. 6 - 148354684 - 148355198 [+] on Build GRCm38 104
          GenBankmRNA AV470717.1 - 1 106 Chr. 6 - 148354703 - 148355151 [+] on Build GRCm38 104
          GenBankmRNA AV490188.1 - 1 141 Chr. 6 - 148354667 - 148355176 [+] on Build GRCm38 104
          GenBankmRNA AV509662.1 - 1 134 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38 104
          GenBankmRNA AV585136.1 - 1 127 Chr. 6 - 148354678 - 148355194 [+] on Build GRCm38 104
          GenBankmRNA AV444821.1 - 1 127 Chr. 6 - 148354691 - 148355191 [+] on Build GRCm38 104
          GenBankmRNA AA230473.1 - 1 76 Chr. 6 - 148354730 - 148355194 [+] on Build GRCm38 104
          GenBankmRNA AV455637.1 - 1 126 Chr. 6 - 148354690 - 148355179 [+] on Build GRCm38 104
          GenBankmRNA AV457756.1 - 1 130 Chr. 6 - 148354673 - 148355144 [+] on Build GRCm38 104
          GenBankmRNA AV496022.1 - 1 121 Chr. 6 - 148354684 - 148355180 [+] on Build GRCm38 104
          GenBankmRNA BI904197.1 - 1 92 Chr. 6 - 148354749 - 148355474 [+] on Build GRCm38 104
          GenBankmRNA BY034424.1 - 1 95 Chr. 6 - 148354710 - 148355085 [+] on Build GRCm38 104
          GenBankmRNA AA073401.1 - 1 84 Chr. 6 - 148354721 - 148355163 [+] on Build GRCm38 104
          GenBankmRNA AV493657.1 - 1 132 Chr. 6 - 148354672 - 148355152 [+] on Build GRCm38 104
          GenBankmRNA AV491618.1 - 1 116 Chr. 6 - 148354702 - 148355197 [+] on Build GRCm38 104
          GenBankmRNA AV499469.1 - 1 136 Chr. 6 - 148354671 - 148355173 [+] on Build GRCm38 104
          GenBankmRNA AV573947.1 - 1 120 Chr. 6 - 148354685 - 148355142 [+] on Build GRCm38 104

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane and tetratricopeptide repeat containing 1 Tmtc1 Arg99,
          BC023818
          Anti-sense Chr. 6 - 148232430 - 148444845 on NCBI Build GRCm38
          Downstream predicted gene 6313 Gm6313 EG622319 251.2kb Anti-sense Chr. 6 - 148606786 - 148614309 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001754.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600013991.mm.1 TC0600001754.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600013991.mm.1 NR_003634.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-x7z22:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0