Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03682596_g1
          See other GM14379 GE Assays ›
          Gene Symbol:
          Gm14379
          Gene Name:
          predicted gene 14379
          Gene Aliases:
          OTTMUSG00000016482
          Chromosome Location:
          Chr.X: 7375925 - 7378042 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03682596_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14379
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026741.1 1 - 2 361 57
          GenBank mRNA AK040575.1 - 1 - 2 175 57
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB636260.1 - 1 - 2 172 Chr. X - 7376114 - 7377014 [+] on Build GRCm38 57

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitin specific peptidase 27,
          X chromosome
          Usp27x Sfc11,
          Usp27
          -95.0 Anti-sense Chr. X - 7372591 - 7375830 on NCBI Build GRCm38
          Downstream RIKEN cDNA 2010204K13 gene 2010204K13Rik 0610010M13Rik,
          Cip7
          33.8kb Anti-sense Chr. X - 7411817 - 7422988 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000057.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36122 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03634760 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14899939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14899938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15054701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14899929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14546576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14644976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32238142 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598288 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598287 - -
          Affymetrix Mouse Clariom S Array PSR0X00000179.mm.1 TC0X00000057.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000179.mm.1 NR_026741.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000179.mm.1 NR_026741.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000182.mm.1 TC0X00000057.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00000182.mm.1 NR_026741.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00000182.mm.1 NR_026741.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix