Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03808589_m1
          See other SOX8 GE Assays ›
          Gene Symbol:
          Sox8
          Gene Name:
          SRY (sex determining region Y)-box 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 25565893 - 25570686 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03808589_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sox8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011447.3 NP_035577.1 1 - 2 579 69
          GenBank mRNA AK032459.1 - 1 - 2 580 69
          AK033460.1 - 1 - 2 580 69
          AK034231.1 - 1 - 2 580 69
          AK046473.1 - 1 - 2 580 69
          AK053716.1 - 1 - 2 580 69
          AK145294.1 - 1 - 2 580 69
          BC085619.1 - 1 - 2 555 6306276 69
          Z18957.1 - 1 - 2 98 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001665.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sox8

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          neural crest cell migration
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          signal transduction
          spermatogenesis
          peripheral nervous system development
          male gonad development
          positive regulation of gene expression
          regulation of hormone levels
          positive regulation of gliogenesis
          positive regulation of osteoblast proliferation
          skeletal muscle cell differentiation
          negative regulation of apoptotic process
          cell fate commitment
          fat cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          negative regulation of photoreceptor cell differentiation
          cell maturation
          enteric nervous system development
          oligodendrocyte differentiation
          Sertoli cell development
          astrocyte fate commitment
          retina development in camera-type eye
          retinal rod cell differentiation
          adipose tissue development
          morphogenesis of a branching epithelium
          renal vesicle induction
          ureter morphogenesis
          metanephric nephron tubule formation
          positive regulation of kidney development
          positive regulation of branching involved in ureteric bud morphogenesis
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          transcription factor binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700015187.mm.1 TC1700001665.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700015187.mm.1 NM_011447.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015187.mm.1 NM_011447.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015184.mm.1 TC1700001665.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700015184.mm.1 NM_011447.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015184.mm.1 NM_011447.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015186.mm.1 TC1700001665.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700015186.mm.1 NM_011447.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015186.mm.1 NM_011447.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 344295 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0