Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929254_s1
          See other 1700008H02RIK GE Assays ›
          Gene Symbol:
          1700008H02Rik
          Gene Name:
          RIKEN cDNA 1700008H02 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929254_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700008H02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005767.1 - 1 1547 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream adhesion G protein-coupled receptor L3 Adgrl3 5430402I23Rik,
          CIRL-3,
          D130075K09Rik,
          Gm1379,
          LEC3,
          Lphn3,
          mKIAA0768
          -291.9kb Anti-sense Chr. 5 - 81021199 - 81827825 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700031L13 gene 1700031L13Rik 1.3kb Sense Chr. 5 - 82122693 - 82124711 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554570 - -
          Whole Mouse Genome G4122A A_51_P248793 AK005767.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04885577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23844202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04346826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432652_PM_at - -
          Mouse Genome 430 2.0 Array 1432652_PM_at - -
          Mouse Genome 430 2.0 Array 1432653_PM_at - -
          Mouse Genome 430 2.0 Array 1432652_at - -
          Mouse Genome 430 2.0 Array 1432653_at - -
          Mouse Expression Set 430 1432653_at - -
          Mouse Expression Set 430 1432652_PM_at - -
          Mouse Expression Set 430 1432653_PM_at - -
          Mouse Expression Set 430 1432652_at - -
          Affymetrix HT Mouse Genome 430B Array 1432652_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432652_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 626019 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104610692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline