Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929290_m1
          See other 1700010N08RIK GE Assays ›
          Gene Symbol:
          1700010N08Rik
          Gene Name:
          RIKEN cDNA 1700010N08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 38379368 - 38386551 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929290_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700010N08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005849.1 - 439 83
          AK076675.1 - 422 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ047057.1 - 2 - 3 285 Chr. 7 - 39317736 - 39354451 [+] on Build GRCm38 83
          GenBankmRNA BB614962.1 - 3 - 4 422 Chr. 7 - 38379383 - 38416868 [+] on Build GRCm38 83
          GenBankmRNA CA464463.1 - 2 - 3 276 Chr. 7 - 38408163 - 38442739 [+] on Build GRCm38 83
          GenBankmRNA BY705994.1 - 439 Chr. - - [-] on Build 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          21276
          Gm21276 -547.9kb Anti-sense Chr. 7 - 38762406 - 38770304 on NCBI Build GRCm38
          Downstream predicted gene 5591 Gm5591 EG434171 101.3kb Anti-sense Chr. 7 - 38518139 - 38528193 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355064 AK076675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355064 AV260173.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355064 AK076675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P355064 AV260173.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355064 AK076675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355064 AV260173.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355064 AK076675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P355064 AV260173.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06170506 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06170496 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06170438 - -
          Whole Mouse Genome G4122A A_52_P355064 AK076675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355064 AV260173.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355064 AK076675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P355064 AV260173.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06170438 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06170496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25066490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25066482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06532927 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430315_PM_at - -
          Mouse Genome 430 2.0 Array 1430315_PM_at - -
          Mouse Genome 430 2.0 Array 1430315_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 545951_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 545951_at - -
          Mouse Expression Set 430 1430315_at - -
          Mouse Expression Set 430 1430315_PM_at - -
          Murine Genome U74v2 Set 166295_PM_f_at - -
          Murine Genome U74v2 Set 166295_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1430315_at - -
          Affymetrix HT Mouse Genome 430B Array 1430315_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106520603 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0