Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929672_s1
          See other 1700058J15RIK GE Assays ›
          Gene Symbol:
          1700058J15Rik
          Gene Name:
          RIKEN cDNA 1700058J15 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929672_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700058J15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK006832.1 - 1 651 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4921508D12 gene 4921508D12Rik -37.2kb Anti-sense Chr. 2 - 132424741 - 132452499 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700026D11 gene 1700026D11Rik 105.0 Anti-sense Chr. 2 - 132490728 - 132524057 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02158947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02158931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02158935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21640604 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 170165_at - -
          Murine Genome U74v2 Set 170165_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 302690 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101770519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0