Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03929825_m1
          See other 1700105G05RIK GE Assays ›
          Gene Symbol:
          1700105G05Rik
          Gene Name:
          RIKEN cDNA 1700105G05 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03929825_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700105G05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007126.1 - 1 - 2 220 194

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          26512
          Gm26512 -157.7kb Sense Chr. 12 - 90738212 - 90812245 on NCBI Build GRCm38
          Downstream centrosomal protein 128 Cep128 4930534B04Rik,
          5430424K18Rik,
          AI449708,
          AI480494,
          E230024F20
          27.0kb Anti-sense Chr. 12 - 90998490 - 91384416 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10140318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10018554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10125810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10111579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09958676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28632336 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 168581_i_at - -
          Murine Genome U74v2 Set 168581_PM_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 427847 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix