Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03930554_s1
          See other 2310033F14RIK GE Assays ›
          Gene Symbol:
          2310033F14Rik Pbxip1
          Gene Name:
          RIKEN cDNA 2310033F14 gene
          pre B cell leukemia transcription factor interacting protein 1
          Gene Aliases:
          -
          4732463H20Rik
          Chromosome Location:
          -
          Chromosome Location:
          Chr.3: 89436675 - 89450952 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm03930554_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310033F14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK009590.1 - 1 47 59
          Gene Symbol: Pbxip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146131.2 NP_666243.1 11 4349 59
          GenBank mRNA CD742482.1 - 1 59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CX203378.1 - 1 40 Chr. 3 - 89450781 - 89450944 [+] on Build GRCm38 59
          GenBankmRNA BM730835.1 - 1 Chr. 3 - 89450458 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA AV220340.2 - 1 486 Chr. 3 - 89450368 - 89450941 [+] on Build GRCm38 59
          GenBankmRNA EH106658.1 - 1 300 Chr. 3 - 89450531 - 89450877 [+] on Build GRCm38 59
          GenBankmRNA BY709265.1 - 1 47 Chr. 3 - 89450774 - 89450941 [+] on Build GRCm38 59
          GenBankmRNA AV228890.1 - 1 166 Chr. 3 - 89450673 - 89450941 [+] on Build GRCm38 59
          GenBankmRNA CF425257.1 - 1 120 Chr. 3 - 89450721 - 89450952 [+] on Build GRCm38 59
          GenBankmRNA AW208779.1 - 1 Chr. 3 - 89450475 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA CD741017.1 - 1 Chr. 3 - 89450179 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA AA240834.1 - 1 Chr. 3 - 89450682 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA AW701924.1 - 1 Chr. 3 - 89450437 - 89450938 [-] on Build GRCm38 59
          GenBankmRNA BP768267.1 - 1 Chr. 3 - 89450380 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA CX210772.1 - 1 314 Chr. 3 - 89450507 - 89450911 [+] on Build GRCm38 59
          GenBankmRNA BG092714.1 - 1 Chr. 3 - 89450475 - 89450952 [-] on Build GRCm38 59
          GenBankmRNA BE950699.1 - 1 Chr. 3 - 89450602 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA AI846081.1 - 1 Chr. 3 - 89450773 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA BM052172.1 - 1 Chr. 3 - 89450458 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA AI427511.1 - 1 Chr. 3 - 89450653 - 89450943 [-] on Build GRCm38 59
          GenBankmRNA CX205623.1 - 1 50 Chr. 3 - 89450771 - 89450944 [+] on Build GRCm38 59
          GenBankmRNA AW107937.1 - 1 Chr. 3 - 89450459 - 89450944 [-] on Build GRCm38 59
          GenBankmRNA BY613220.1 - 1 284 Chr. 3 - 89450540 - 89450941 [+] on Build GRCm38 59
          GenBankmRNA BF584079.1 - 1 250 Chr. 3 - 89450583 - 89450944 [+] on Build GRCm38 59
          GenBankmRNA BB678582.1 - 1 302 Chr. 3 - 89450519 - 89450941 [+] on Build GRCm38 59
          GenBankmRNA BY606286.1 - 1 244 Chr. 3 - 89450577 - 89450943 [+] on Build GRCm38 59
          GenBankmRNA CV677237.1 - 1 Chr. 3 - 89450404 - 89450902 [-] on Build GRCm38 59
          GenBankmRNA CK382816.1 - 1 120 Chr. 3 - 89450721 - 89450952 [+] on Build GRCm38 59
          GenBankmRNA AW146195.1 - 1 Chr. 3 - 89450393 - 89450932 [-] on Build GRCm38 59
          GenBankmRNA EH095564.1 - 1 410 Chr. 3 - 89450411 - 89450877 [+] on Build GRCm38 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream pygopus 2 Pygo2 1190004M21Rik -15.6kb Sense Chr. 3 - 89430132 - 89435131 on NCBI Build GRCm38
          Downstream phosphomevalonate kinase Pmvk 1110011E12Rik,
          2900002L22Rik,
          AV001327,
          PMK,
          PMKA,
          PMKASE
          3.6kb Sense Chr. 3 - 89454546 - 89469014 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000765.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310033F14Rik
          Click here to find siRNAs for Gene Pbxip1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1460344_PM_at - -
          Mouse Genome 430 2.0 Array 1460344_at - -
          Affymetrix Mouse Clariom S Array PSR0300005821.mm.1 TC0300000765.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300005821.mm.1 NM_146131.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005821.mm.1 NM_146131.2 Refseq
          Mouse Expression Set 430 1460344_PM_at - -
          Mouse Expression Set 430 1460344_at - -
          Mouse Genome 430A 2.0 Array 1460344_PM_at - -
          Mouse Genome 430A 2.0 Array 1460344_at - -
          Affymetrix HT Mouse Genome 430A Array 1460344_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460344_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix