Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130118H10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021395.1 | - | 1 | 45 | 58 | |
AK165039.1 | - | 1 | 5209 | 58 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | expressed sequence AI839979 | AI839979 | -991.0 | Anti-sense | Chr. 5 - 31569592 - 31571397 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 38424 |
Gm38424 | 22.7kb | Sense | Chr. 5 - 31595752 - 31663432 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |