Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03933207_m1
          See other A730085K08RIK GE Assays ›
          Gene Symbol:
          A730085K08Rik
          Gene Name:
          RIKEN cDNA A730085K08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 122382022 - 122398801 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03933207_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A730085K08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045967.1 2 - 3 225 94
          GenBank mRNA AK033049.1 - 2 - 3 223 94
          AK043328.1 - 226 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB624012.1 - 2 - 3 223 Chr. 9 - 122382064 - 122398798 [-] on Build GRCm38 94
          GenBankmRNA AV360585.2 - 2 - 3 235 Chr. 9 - 122382066 - 122398798 [-] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream abhydrolase domain containing 5 Abhd5 1300003D03Rik,
          2010002J10Rik,
          CDS,
          CGI-58,
          IECN5,
          NCIE2
          -542.0 Anti-sense Chr. 9 - 122351539 - 122381524 on NCBI Build GRCm38
          Downstream RIKEN cDNA 9530059O14 gene 9530059O14Rik 182.9kb Anti-sense Chr. 9 - 122572501 - 122580647 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003287.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450451 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410823 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307764 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14417 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127867 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127867 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06954185 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119706 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08064754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07554441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26889196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07554453 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900027709.mm.1 TC0900003287.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027709.mm.1 NR_045967.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027709.mm.1 NR_045967.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887228 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0