Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03934876_g1
          See other 9530082P21RIK GE Assays ›
          Gene Symbol:
          9530082P21Rik
          Gene Name:
          RIKEN cDNA 9530082P21 gene
          Gene Aliases:
          E030034P13Rik
          Chromosome Location:
          Chr.17: 23749236 - 23754065 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03934876_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530082P21Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015472.1 1 - 2 474 56
          GenBank mRNA AK037113.1 - 683 56
          AK079285.1 - 2 - 3 269 56
          AK087205.1 - 2 - 3 303 56
          AK087218.1 - 1 - 2 475 56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB664356.1 - 1 - 2 475 Chr. 17 - 23749236 - 23749987 [+] on Build GRCm38 56
          GenBankmRNA BY722295.1 - 2 - 3 269 Chr. 17 - 23749370 - 23750185 [+] on Build GRCm38 56
          GenBankmRNA BB664336.1 - 2 - 3 303 Chr. 17 - 23749337 - 23750135 [+] on Build GRCm38 56
          GenBankmRNA CA493465.1 - 1 - 2 378 Chr. 17 - 23749369 - 23750156 [+] on Build GRCm38 56

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream kringle containing transmembrane protein 2 Kremen2 2900054E04Rik,
          Krm2
          -3.5kb Anti-sense Chr. 17 - 23741199 - 23745829 on NCBI Build GRCm38
          Downstream FLYWCH-type zinc finger 1 Flywch1 E030034P13Rik,
          mKIAA1552
          2.6kb Anti-sense Chr. 17 - 23752797 - 23771591 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000319.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2470474 AK087205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2470474 AK079285.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768369 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 AB029501.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 AK037699.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 AK054475.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 AK136799.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 AK165334.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 BC049189.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 BC062131.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764168 NM_172588.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40430 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137768 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03292276 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08737536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03292287 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136077 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08737536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441996_PM_at - -
          Mouse Genome 430 2.0 Array 1441996_at - -
          Mouse Genome 430 2.0 Array 1441996_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 638247_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 638247_at - -
          Affymetrix Mouse Clariom S Array PSR1700002327.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002327.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002327.mm.1 NR_015472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002326.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002326.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002326.mm.1 NR_015472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002328.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002328.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002328.mm.1 NR_015472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002329.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002329.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002329.mm.1 NR_015472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002325.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002325.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002325.mm.1 NR_015472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002333.mm.1 TC1700000319.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002333.mm.1 NR_015472.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002333.mm.1 NR_015472.1 Refseq
          Mouse Expression Set 430 1441996_PM_at - -
          Mouse Expression Set 430 1441996_at - -
          Affymetrix HT Mouse Genome 430B Array 1441996_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441996_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 546003 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0