Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03935585_m1
          See other GM2721 GE Assays ›
          Gene Symbol:
          Gm2721
          Gene Name:
          predicted gene 2721
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 105394856 - 105403760 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03935585_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm2721
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045085.1 2 - 3 135 98
          GenBank mRNA AK039118.1 - 2 - 3 136 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB632951.1 - 2 - 3 136 Chr. 12 - 105395723 - 105403760 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Tcl1 upstream neural differentiation associated RNA Tunar 2810011L19Rik,
          Tuna
          -10.9kb Anti-sense Chr. 12 - 105336594 - 105383932 on NCBI Build GRCm38
          Downstream RIKEN cDNA D430019H16 gene D430019H16Rik AI852661 55.8kb Anti-sense Chr. 12 - 105453856 - 105493095 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002307.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973993 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10374002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10175307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10175312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09894842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10055499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10373989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09861281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09917863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10245020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10175315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10004804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28708986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10373990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10374003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09751728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10424807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10055498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10373999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09900968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28709031 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200016100.mm.1 TC1200002307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016100.mm.1 NR_045085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016100.mm.1 NR_045085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016101.mm.1 TC1200002307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016101.mm.1 NR_045085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016101.mm.1 NR_045085.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016099.mm.1 TC1200002307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200016099.mm.1 NR_045085.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200016099.mm.1 NR_045085.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 386449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0