Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03936535_g1
          See other GM14317 GE Assays ›
          Gene Symbol:
          Gm14317
          Gene Name:
          predicted gene 14317
          Gene Aliases:
          OTTMUSG00000016294
          Chromosome Location:
          Chr.2: 164549485 - 164559748 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03936535_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14317
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK043662.1 - 2 - 3 393 113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BU524700.1 - 2 - 3 300 Chr. 2 - 164550622 - 164558877 [+] on Build GRCm38 113
          GenBankmRNA BB641322.1 - 2 - 3 393 Chr. 2 - 164550530 - 164558942 [+] on Build GRCm38 113
          GenBankmRNA BY270849.1 - 2 - 3 311 Chr. 2 - 164550612 - 164558764 [+] on Build GRCm38 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphatidylinositol glycan anchor biosynthesis,
          class T
          Pigt 2510012P17Rik,
          4930534E15Rik,
          CGI-06,
          NDAP,
          Ndap7
          -49.8kb Sense Chr. 2 - 164497511 - 164508301 on NCBI Build GRCm38
          Downstream WAP four-disulfide core domain 2 Wfdc2 1600023A02Rik,
          HE4,
          WAP5
          2.8kb Sense Chr. 2 - 164562563 - 164568510 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P117484 AK043662.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 326180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416368 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P117484 AK043662.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04772379 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117484 AK043662.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P01800859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01896992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01713598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01713595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02043278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01800860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01320826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01713599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818481 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21818503 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0