Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C230021G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045502.1 | - | 1 - 2 | 243 | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | coiled-coil domain containing 59 | Ccdc59 |
2300004H16Rik, D10Ertd718e |
-620.3kb | Anti-sense | Chr. 10 - 105841479 - 105847510 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 4930532I03 gene | 4930532I03Rik | 84.5kb | Sense | Chr. 10 - 106555509 - 106606402 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |