Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03938875_s1
          See other 4930477J04RIK GE Assays ›
          Gene Symbol:
          4930477J04Rik
          Gene Name:
          RIKEN cDNA 4930477J04 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03938875_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930477J04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015580.1 - 1 448 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715130.1 - 1 448 Chr. 18 - 36320906 - 36321492 [-] on Build GRCm38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream IgA inducing protein Igip 6330403M23Rik -19.4kb Anti-sense Chr. 18 - 36300101 - 36301478 on NCBI Build GRCm38
          Downstream cysteine-rich transmembrane module containing 1 Cystm1 0610010O12Rik 27.1kb Anti-sense Chr. 18 - 36348624 - 36393370 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08965852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13887882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14039535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13846478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13865507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623246 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13830588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13618729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13655653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13963618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14082234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13846463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13846475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13655638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13655639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13840698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14082235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13835885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31623309 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101780538 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix