Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03939038_g1
          See other 4930511J24RIK GE Assays ›
          Gene Symbol:
          4930511J24Rik
          Gene Name:
          RIKEN cDNA 4930511J24 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03939038_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930511J24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015759.1 - 1 - 2 151 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715302.1 - 1 - 2 151 Chr. 12 - 109911785 - 109912707 [+] on Build GRCm38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream miRNA containing gene Mirg 2810474H01Rik,
          AI852992,
          Gm2928,
          Meg9,
          mir-410
          -162.3kb Sense Chr. 12 - 109734981 - 109749458 on NCBI Build GRCm38
          Downstream RIKEN cDNA 3110009F21 gene 3110009F21Rik 238.1kb Anti-sense Chr. 12 - 110151654 - 110158073 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2406802 AK015759.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121022 - -
          Whole Mouse Genome G4122A A_52_P32393 AK015759.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10129469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09863260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10227618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09863274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28735480 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10129475 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431565_PM_at - -
          Mouse Genome 430 2.0 Array 1431565_at - -
          Mouse Genome 430 2.0 Array 1431565_PM_at - -
          Mouse Expression Set 430 1431565_PM_at - -
          Mouse Expression Set 430 1431565_at - -
          Affymetrix HT Mouse Genome 430B Array 1431565_at - -
          Affymetrix HT Mouse Genome 430B Array 1431565_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 454473 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103170142 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0