Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1433039_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433038_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433039_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433039_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1433038_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200006997.mm.1 |
TC0200000867.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006997.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006997.mm.1 |
NR_040633.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006998.mm.1 |
TC0200000867.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006998.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006998.mm.1 |
NR_040633.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006996.mm.1 |
TC0200000867.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006996.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006996.mm.1 |
NR_040633.1 |
Refseq |
Mouse Expression Set 430 |
1433039_at |
- |
- |
Mouse Expression Set 430 |
1433038_PM_at |
- |
- |
Mouse Expression Set 430 |
1433039_PM_at |
- |
- |
Mouse Expression Set 430 |
1433038_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1433039_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1433039_PM_at |
- |
- |