Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03941086_s1
          See other 5730405A17RIK GE Assays ›
          Gene Symbol:
          5730405A17Rik
          Gene Name:
          RIKEN cDNA 5730405A17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03941086_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 5730405A17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK017489.1 - 1 1249 117

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream SH3 domain containing ring finger 1 Sh3rf1 2200003J05Rik,
          Posh,
          R75531,
          Sh3md2
          Anti-sense Chr. 8 - 61224171 - 61464778 on NCBI Build GRCm38
          Downstream carbonyl reductase 4 Cbr4 A730083J17Rik,
          BC009118
          69.7kb Anti-sense Chr. 8 - 61487734 - 61503500 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2280316 AK017489.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 AB007143.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 AK035171.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 AK158299.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 AK188664.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 BC012210.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 BC023238.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 BC125443.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312891 NM_007828.1 refseq_mrna_mouse_NM
          Agilent Six_ChIP_02_of_03_v2 A_68_P07132777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07132431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07048844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07099149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06786467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07303378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06720045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07048840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07048836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07048835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06990617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06843554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06938177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25909824 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432703_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432704_PM_at - -
          Mouse Genome 430 2.0 Array 1432703_PM_at - -
          Mouse Genome 430 2.0 Array 1432704_at - -
          Mouse Genome 430 2.0 Array 1432704_PM_at - -
          Mouse Genome 430 2.0 Array 1432703_at - -
          Mouse Expression Set 430 1432704_PM_at - -
          Mouse Expression Set 430 1432703_PM_at - -
          Mouse Expression Set 430 1432704_at - -
          Mouse Expression Set 430 1432703_at - -
          Affymetrix HT Mouse Genome 430B Array 1432703_at - -
          Affymetrix HT Mouse Genome 430B Array 1432704_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432703_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432704_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0