Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03943659_s1
          See other SH3GLB1 GE Assays ›
          Gene Symbol:
          Sh3glb1
          Gene Name:
          SH3-domain GRB2-like B1 (endophilin)
          Gene Aliases:
          AA409932, AI314629, AU015566, Bif-1, mKIAA0491
          Chromosome Location:
          Chr.3: 144683678 - 144720398 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03943659_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sh3glb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282037.1 NP_001268966.1 11 5523 100
          NM_019464.3 NP_062337.1 9 5460 100
          GenBank mRNA AK052668.1 - 1 1954 100
          AK147816.1 - 1 1931 100
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB499022.2 - 1 559 Chr. 3 - 144687217 - 144687903 [-] on Build GRCm38 100
          GenBankmRNA BM937674.1 - 1 Chr. 3 - 144686911 - 144687557 [+] on Build GRCm38 100
          GenBankmRNA CF586322.1 - 1 Chr. 3 - 144686911 - 144687799 [+] on Build GRCm38 100
          GenBankmRNA CB194902.1 - 1 727 Chr. 3 - 144687235 - 144688066 [-] on Build GRCm38 100
          GenBankmRNA AI746369.1 - 1 Chr. 3 - 144687159 - 144687567 [+] on Build GRCm38 100
          GenBankmRNA AW412578.1 - 1 Chr. 3 - 144687215 - 144687721 [+] on Build GRCm38 100
          GenBankmRNA CX219861.1 - 1 663 Chr. 3 - 144687216 - 144688007 [-] on Build GRCm38 100
          GenBankmRNA AA870563.1 - 1 202 Chr. 3 - 144687215 - 144687537 [-] on Build GRCm38 100
          GenBankmRNA BE654756.1 - 1 291 Chr. 3 - 144687161 - 144687635 [-] on Build GRCm38 100
          GenBankmRNA BE334985.1 - 1 502 Chr. 3 - 144687300 - 144687847 [-] on Build GRCm38 100
          GenBankmRNA DV065875.1 - 138 Chr. - - [-] on Build 100
          GenBankmRNA BP768911.1 - 1 Chr. 3 - 144686911 - 144687423 [+] on Build GRCm38 100
          GenBankmRNA AI450300.1 - 1 Chr. 3 - 144687214 - 144687576 [+] on Build GRCm38 100
          GenBankmRNA BB221842.2 - 1 258 Chr. 3 - 144686918 - 144687601 [-] on Build GRCm38 100
          GenBankmRNA AI591655.1 - 1 204 Chr. 3 - 144687082 - 144687537 [-] on Build GRCm38 100
          GenBankmRNA BE457494.1 - 1 Chr. 3 - 144686911 - 144687567 [+] on Build GRCm38 100
          GenBankmRNA BG145796.1 - 1 448 Chr. 3 - 144687268 - 144687792 [-] on Build GRCm38 100
          GenBankmRNA AA014754.1 - 487 Chr. - - [-] on Build 100
          GenBankmRNA CX205176.1 - 1 155 Chr. 3 - 144686911 - 144687499 [-] on Build GRCm38 100
          GenBankmRNA CR518735.1 - 1 Chr. 3 - 144687210 - 144687449 [+] on Build GRCm38 100

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream selenoprotein Sep15 9430015P09Rik -89.5kb Anti-sense Chr. 3 - 144570427 - 144597677 on NCBI Build GRCm38
          Downstream chloride channel calcium activated 3A1 Clca3a1 Cacc,
          Clca1
          40.4kb Sense Chr. 3 - 144729697 - 144760972 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003077.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sh3glb1

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidic acid biosynthetic process
          apoptotic process
          mitochondrion organization
          phospholipid biosynthetic process
          positive regulation of autophagy
          regulation of protein stability
          positive regulation of protein oligomerization
          regulation of cytokinesis
          receptor catabolic process
          cellular response to amino acid starvation
          cellular response to glucose starvation
          positive regulation of phosphatidylinositol 3-kinase activity
          autophagic cell death
          'de novo' posttranslational protein folding
          protein complex oligomerization
          membrane fission
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          positive regulation of membrane tubulation
          protein localization to vacuolar membrane
          positive regulation of protein targeting to mitochondrion
          positive regulation of autophagosome assembly
          fatty acid binding
          protein binding
          lipid binding
          lysophosphatidic acid acyltransferase activity
          identical protein binding
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300024149.mm.1 TC0300003077.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024149.mm.1 NM_001282037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024149.mm.1 NM_019464.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024148.mm.1 TC0300003077.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024148.mm.1 NM_001282037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024148.mm.1 NM_019464.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024150.mm.1 TC0300003077.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024150.mm.1 NM_001282037.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024150.mm.1 NM_019464.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 633640 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0