Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03943748_s1
          See other KDM5C GE Assays ›
          Gene Symbol:
          Kdm5c
          Gene Name:
          lysine (K)-specific demethylase 5C
          Gene Aliases:
          D930009K15Rik, Jarid1c, Smcx, mKIAA0234
          Chromosome Location:
          Chr.X: 152233196 - 152279102 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03943748_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kdm5c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013668.4 NP_038696.2 26 10576 94
          GenBank mRNA AK052994.1 - 1 1244 94
          AK148627.1 - 1 2236 94
          BG075333.2 - 1 94
          BQ177188.1 - 1 94
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BM932394.1 - 1 Chr. X - 152278332 - 152278936 [-] on Build GRCm38 94
          GenBankmRNA BY407362.1 - 1 53 Chr. X - 152278608 - 152278939 [+] on Build GRCm38 94
          GenBankmRNA AW240860.1 - 1 Chr. X - 152278514 - 152278932 [-] on Build GRCm38 94
          GenBankmRNA C88261.1 - Chr. - - [-] on Build 94
          GenBankmRNA AW549557.2 - 1 Chr. X - 152278471 - 152278932 [-] on Build GRCm38 94
          GenBankmRNA BB522751.2 - 1 372 Chr. X - 152278288 - 152278921 [+] on Build GRCm38 94
          GenBankmRNA AW122702.1 - 1 Chr. X - 152278460 - 152278940 [-] on Build GRCm38 94
          GenBankmRNA AW555812.1 - 1 Chr. X - 152278237 - 152278932 [-] on Build GRCm38 94
          GenBankmRNA BE648268.1 - 1 586 Chr. X - 152278077 - 152278799 [+] on Build GRCm38 94
          GenBankmRNA BB667285.1 - 1 188 Chr. X - 152278475 - 152279097 [+] on Build GRCm38 94

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream IQ motif and Sec7 domain 2 Iqsec2 Brag1,
          mKIAA0522
          -52.2kb Sense Chr. X - 152144183 - 152225237 on NCBI Build GRCm38
          Downstream Kdm5c adjacent non-coding transcript Kantr 2810454L23Rik,
          2900056M20Rik,
          Spasm
          15.9kb Anti-sense Chr. X - 152294824 - 152327493 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kdm5c

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          histone H3-K4 demethylation
          regulation of circadian rhythm
          negative regulation of transcription, DNA-templated
          rhythmic process
          sulfonate dioxygenase activity
          DNA binding
          zinc ion binding
          oxidoreductase activity
          2-oxoglutarate-dependent dioxygenase activity
          2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
          procollagen-proline dioxygenase activity
          histone H3-methyl-lysine-4 demethylase activity
          hypophosphite dioxygenase activity
          identical protein binding
          DNA-N1-methyladenine dioxygenase activity
          metal ion binding
          dioxygenase activity
          C-19 gibberellin 2-beta-dioxygenase activity
          C-20 gibberellin 2-beta-dioxygenase activity

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227553 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162423 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162423 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1435348_PM_at - -
          Mouse Genome 430 2.0 Array 1435348_PM_at - -
          Mouse Genome 430 2.0 Array 1435348_at - -
          Mouse Expression Set 430 1435348_PM_at - -
          Mouse Expression Set 430 1435348_at - -
          Affymetrix HT Mouse Genome 430B Array 1435348_at - -
          Affymetrix HT Mouse Genome 430B Array 1435348_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0