Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03944581_s1
          See other 9430029E18RIK GE Assays ›
          Gene Symbol:
          9430029E18Rik
          Gene Name:
          RIKEN cDNA 9430029E18 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03944581_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430029E18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK034727.1 - 1 2980 92
          AK077816.1 - 1 2449 92

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream T cell lymphoma invasion and metastasis 1 Tiam1 AI847750,
          D16Ium10,
          D16Ium10e
          Sense Chr. 16 - 89787111 - 90145483 on NCBI Build GRCm38
          Downstream predicted gene 10789 Gm10789 ENSMUSG00000074924 23.2kb Anti-sense Chr. 16 - 90142237 - 90146827 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12590279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12956921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955098 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30955100 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1441381_PM_at - -
          Mouse Genome 430 2.0 Array 1441381_at - -
          Mouse Genome 430 2.0 Array 1441381_PM_at - -
          Mouse Expression Set 430 1441381_PM_at - -
          Mouse Expression Set 430 1441381_at - -
          Affymetrix HT Mouse Genome 430B Array 1441381_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441381_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0