Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03947851_s1
          See other GM10657 GE Assays ›
          Gene Symbol:
          Gm10657
          Gene Name:
          predicted gene 10657
          Gene Aliases:
          ENSMUSG00000074273
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03947851_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10657
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK132138.1 - 1 951 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB321112.1 - 1 525 Chr. 9 - 58555224 - 58555847 [+] on Build GRCm38 82
          GenBankmRNA CK625627.1 - 1 Chr. 9 - 58555248 - 58555848 [-] on Build GRCm38 82
          GenBankmRNA BF461211.1 - 1 Chr. 9 - 58555555 - 58555849 [-] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CD276 antigen Cd276 6030411F23Rik,
          AU016588,
          B7RP-2,
          B7h3
          Anti-sense Chr. 9 - 58524300 - 58555033 on NCBI Build GRCm38
          Downstream neuroplastin Nptn AW554172,
          SDR-1,
          Sdfr1
          26.4kb Sense Chr. 9 - 58582240 - 58652879 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177347 AK132138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177347 AK132138.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07484549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07553702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07599450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07986162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08104559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07721979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07592522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07700494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07553703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07592526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07673055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07492950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07986167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26548583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix