Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830418P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040466.1 | 2 - 3 | 290 | 95 | ||
GenBank mRNA | AK079871.1 | - | 1 - 2 | 143 | 95 | |
AK134047.1 | - | 2 - 3 | 291 | 95 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
beaded filament structural protein 2, phakinin |
Bfsp2 |
AI448593, CP49 |
Anti-sense | Chr. 9 - 103424924 - 103480414 on NCBI Build GRCm38 | |
Downstream | predicted gene 16252 | Gm16252 | OTTMUSG00000032743 | 44.7kb | Sense | Chr. 9 - 103472049 - 103503357 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |