Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948339_s1
          See other AI450353 GE Assays ›
          Gene Symbol:
          AI450353
          Gene Name:
          expressed sequence AI450353
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 83293430 - 83294929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948339_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AI450353
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028364.1 1 855 83
          GenBank mRNA AK134743.1 - 1 855 83
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY595740.1 - 1 125 Chr. 11 - 83294160 - 83294627 [+] on Build GRCm38 83
          GenBankmRNA BF139955.1 - 1 107 Chr. 11 - 83294177 - 83294624 [+] on Build GRCm38 82
          GenBankmRNA BX517310.1 - 1 366 Chr. 11 - 83293919 - 83294426 [+] on Build GRCm38 83
          GenBankmRNA AA138581.1 - 1 335 Chr. 11 - 83293950 - 83294468 [+] on Build GRCm38 83
          GenBankmRNA CF381692.1 - 1 Chr. 11 - 83293818 - 83294384 [-] on Build GRCm38 83
          GenBankmRNA BY590120.1 - 1 116 Chr. 11 - 83294169 - 83294626 [+] on Build GRCm38 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          C/D box 7
          Snord7 Sense Chr. 11 - 83294304 - 83294399 on NCBI Build GRCm38
          Downstream adaptor-related protein complex 2,
          beta 1 subunit
          Ap2b1 1300012O03Rik,
          AI788979
          4.0kb Sense Chr. 11 - 83298969 - 83405035 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001246.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1440345_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379652 - -
          Mouse Genome 430 2.0 Array 1440345_at - -
          Mouse Genome 430 2.0 Array 1440345_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100011697.mm.1 TC1100001246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011697.mm.1 NR_028364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011697.mm.1 NR_028364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011688.mm.1 TC1100001246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011688.mm.1 NR_028364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011688.mm.1 NR_028364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011691.mm.1 TC1100001246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011691.mm.1 NR_028364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011691.mm.1 NR_028364.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011689.mm.1 TC1100001246.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100011689.mm.1 NR_028362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011689.mm.1 NR_028362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011689.mm.1 NR_028364.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100011689.mm.1 NR_028364.1 Refseq
          Mouse Expression Set 430 1440345_PM_at - -
          Mouse Expression Set 430 1440345_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix