Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948363_g1
          See other GM10125 GE Assays ›
          Gene Symbol:
          Gm10125
          Gene Name:
          predicted gene 10125
          Gene Aliases:
          ENSMUSG00000063087, Zeb1os
          Chromosome Location:
          Chr.18: 5491501 - 5592437 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948363_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10125
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033552.1 2 - 3 359 71
          GenBank mRNA AK134789.1 - 2 - 3 316 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY727378.1 - 2 - 3 361 Chr. 18 - 5567426 - 5592439 [-] on Build GRCm38 71
          GenBankmRNA BY718683.1 - 2 - 3 309 Chr. 18 - 5525218 - 5592387 [-] on Build GRCm38 71
          GenBankmRNA AW048493.1 - 2 - 3 112 Chr. 18 - 5575278 - 5592190 [-] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 438 Zfp438 9430031G09,
          9430091M14Rik,
          B830013J05Rik
          -248.1kb Sense Chr. 18 - 5210027 - 5334962 on NCBI Build GRCm38
          Downstream zinc finger E-box binding homeobox 1 Zeb1 3110032K11Rik,
          AREB6,
          BZP,
          MEB1,
          Nil2,
          TCF-8,
          Tcf18,
          Tcf8,
          Tw,
          ZEB,
          Zfhep,
          Zfhx1a,
          Zfx1a,
          Zfx1ha,
          [delta]EF1
          8.1kb Anti-sense Chr. 18 - 5591860 - 5775468 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000937.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P139032 AK046967.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2337970 AK138232.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967494 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64160 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87891 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140857 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130616 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124540 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P139032 AK046967.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900726 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03403868 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130616 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140857 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900775 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900670 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900871 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03403921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03403827 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08900627 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124540 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139032 AK046967.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130616 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124540 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140857 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08900652 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03403827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13981530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13962315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13980606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13851602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14013354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13962306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13962311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31468992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14032048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31469273 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10457248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457244 - -
          Affymetrix Mouse Gene 1.1 ST Array 10457243 - -
          Affymetrix Mouse Clariom S Array PSR1800006842.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006842.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006842.mm.1 NR_033552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006838.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006838.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006838.mm.1 NR_033552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006843.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006843.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006843.mm.1 NR_033552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006855.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006855.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006855.mm.1 NR_033552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006840.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006840.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006840.mm.1 NR_033552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006848.mm.1 TC1800000937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800006848.mm.1 NR_033552.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800006848.mm.1 NR_033552.1 Refseq
          Mouse Expression Set 430 1441786_at - -
          Mouse Expression Set 430 1441786_PM_at - -
          Murine Genome U74v2 Set 134522_PM_at - -
          Murine Genome U74v2 Set 138168_PM_at - -
          Murine Genome U74v2 Set 138168_at - -
          Murine Genome U74v2 Set 134522_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 103140097 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2565835 - -
          Illumina Sentrix mouse-6.v1 bead array 103140097 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103140097 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2565835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0