Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948596_m1
          See other AU019990 GE Assays ›
          Gene Symbol:
          AU019990
          Gene Name:
          expressed sequence AU019990
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 132598196 - 132653067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948596_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AU019990
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033469.1 3 - 4 608 101
          GenBank mRNA AK135901.1 - 3 - 4 610 101

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae) Gpcpd1 2310004G06Rik,
          2310032D16Rik,
          AU015213,
          Gde5,
          Prei4,
          mKIAA1434
          -69.7kb Anti-sense Chr. 2 - 132529082 - 132578282 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1110034G24 gene 1110034G24Rik 36.7kb Sense Chr. 2 - 132686875 - 132751055 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002037.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2204779 AK135901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2204779 BB729657.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2204779 BG069619.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2204779 NR_033469.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082449 AK166556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082449 NR_033468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2204779 AK135901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2204779 BB729657.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2204779 BG069619.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2204779 NR_033469.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082449 AK166556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082449 NR_033468.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04697475 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04697500 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04697448 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00496568 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00496568 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04697475 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02274512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01827259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02274515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01872948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01766527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01227348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01766528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01956278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01372458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01200913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01372461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01309684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01814830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01592977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01446219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02108405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01538874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01974040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01827260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01206374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02004190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02225076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01538872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01235512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01610263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21641241 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.1 ST Array 10476347 - -
          Affymetrix Mouse Clariom S Array PSR0200016222.mm.1 TC0200002037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016222.mm.1 NR_033468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016222.mm.1 NR_033468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016222.mm.1 NR_033469.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016222.mm.1 NR_033469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016227.mm.1 TC0200002037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016227.mm.1 NR_033469.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016227.mm.1 NR_033469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016228.mm.1 TC0200002037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016228.mm.1 NR_033469.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016228.mm.1 NR_033469.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016221.mm.1 TC0200002037.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200016221.mm.1 NR_033469.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200016221.mm.1 NR_033469.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline