Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03948874_m1
          See other GM13446 GE Assays ›
          Gene Symbol:
          Gm13446
          Gene Name:
          predicted gene 13446
          Gene Aliases:
          OTTMUSG00000012261
          Chromosome Location:
          Chr.2: 35549534 - 35558702 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03948874_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13446
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045894.1 3 - 4 1158 78
          GenBank mRNA AK138201.1 - 3 - 4 1159 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glycoprotein galactosyltransferase alpha 1,
          3
          Ggta1 AW108479,
          GALT,
          Gal,
          Ggta,
          Ggta-1,
          alpha Gal,
          alpha3GalT
          -85.2kb Sense Chr. 2 - 35400179 - 35464381 on NCBI Build GRCm38
          Downstream disabled 2 interacting protein Dab2ip 2310011D08Rik,
          AI480459,
          Aip1,
          mKIAA1743
          3.3kb Anti-sense Chr. 2 - 35558458 - 35730994 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003372.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 AK005491.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 AK167563.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 AK171850.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 BC064768.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 NM_011452.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853139 U96705.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84381 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166713 AK138201.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00343832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1439603_PM_at - -
          Mouse Genome 430 2.0 Array 1439603_at - -
          Mouse Genome 430 2.0 Array 1439603_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200028993.mm.1 TC0200003372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028993.mm.1 NR_045894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028993.mm.1 NR_045894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028992.mm.1 TC0200003372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028992.mm.1 NR_045894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028992.mm.1 NR_045894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028991.mm.1 TC0200003372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028991.mm.1 NR_045894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028991.mm.1 NR_045894.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028994.mm.1 TC0200003372.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200028994.mm.1 NR_045894.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200028994.mm.1 NR_045894.1 Refseq
          Mouse Expression Set 430 1439603_PM_at - -
          Mouse Expression Set 430 1439603_at - -
          Murine Genome U74v2 Set 128886_PM_at - -
          Murine Genome U74v2 Set 128886_at - -
          Affymetrix HT Mouse Genome 430B Array 1439603_at - -
          Affymetrix HT Mouse Genome 430B Array 1439603_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0