Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949136_m1
          See other GM29687 GE Assays ›
          Gene Symbol:
          Gm29687
          Gene Name:
          predicted gene, 29687
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 26091962 - 26210505 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949136_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm29687
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK139823.1 - 4 - 5 628 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC328066 LOC328066 C920021A13 -620.1kb Anti-sense Chr. 12 - 25316852 - 25471867 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930480M12 gene 4930480M12Rik 113.8kb Anti-sense Chr. 12 - 26211267 - 26240707 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626454 AJ968954.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626454 AK031911.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626454 AK143755.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626454 BC049796.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626454 NM_181423.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445627 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 957770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 AK009237.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 AK011085.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 AK013011.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 AK135332.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 BC014799.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 BC021935.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 920342 NM_025509.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355031 AK043936.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355031 AK139458.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 595018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 662616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157088 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64696 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565498 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02475360 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565456 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02475298 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07565432 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07565540 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07565456 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02475360 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02475298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10101148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10051903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09882407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09800976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10297730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09937794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10341329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09766978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10255001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10401323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10004140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10341384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10197493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09856583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09800971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09892213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10341380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10051897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09769597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09769602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09841240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09967749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10297727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10011416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10341327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10197491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10358031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09841225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09889888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10243370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09769601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10243364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10051898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10051901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10334845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10094415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09921713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10243374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10341331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10004136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09765077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09764124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28299523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28300186 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix