Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03949725_m1
          See other GM6634 GE Assays ›
          Gene Symbol:
          Gm6634
          Gene Name:
          predicted gene 6634
          Gene Aliases:
          EG625901
          Chromosome Location:
          Chr.3: 70772379 - 70807291 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03949725_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6634
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040556.1 1 - 2 70 87
          GenBank mRNA AK144724.1 - 1 - 2 71 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY344100.1 - 70 Chr. - - [-] on Build 87
          GenBankmRNA BY343940.1 - 70 Chr. - - [-] on Build 87
          GenBankmRNA BY344056.1 - 70 Chr. - - [-] on Build 87
          GenBankmRNA BY344456.1 - 70 Chr. - - [-] on Build 87
          GenBankmRNA BY346967.1 - 70 Chr. - - [-] on Build 87
          GenBankmRNA BY734237.1 - 1 - 2 71 Chr. 3 - 70773527 - 70807291 [-] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream otolin 1 homolog (zebrafish) Otol1 Gm414 -769.3kb Anti-sense Chr. 3 - 70007613 - 70028708 on NCBI Build GRCm38
          Downstream sucrase isomaltase (alpha-glucosidase) Sis 2010204N08Rik,
          SI,
          Si-s
          2.1mb Sense Chr. 3 - 72888554 - 72967864 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002147.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2089997 AK144724.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82881 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2089997 AK144724.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04944622 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04944576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04944581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04944576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02568857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02334686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03114182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03193569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03016714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03133365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03133375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02334687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02515318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22227573 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300016137.mm.1 TC0300002147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016137.mm.1 NR_040556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016137.mm.1 NR_040556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016139.mm.1 TC0300002147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016139.mm.1 NR_040556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016139.mm.1 NR_040556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016136.mm.1 TC0300002147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016136.mm.1 NR_040556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016136.mm.1 NR_040556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016138.mm.1 TC0300002147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300016138.mm.1 NR_040556.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300016138.mm.1 NR_040556.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-777976674b-tw4cl:80/100.66.134.228:80.
          git-commit: 6354fc3e62d97773410f28d6123c879be4bd0e1c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0