Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03950542_m1
          See other GM830 GE Assays ›
          Gene Symbol:
          Gm830
          Gene Name:
          predicted gene 830
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 95466913 - 95487394 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03950542_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm830
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK157456.1 - 331 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY752306.1 - 2 - 3 328 Chr. 4 - 95467547 - 95487392 [-] on Build GRCm38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene 12708 Gm12708 OTTMUSG00000008048 -257.8kb Anti-sense Chr. 4 - 95193274 - 95209724 on NCBI Build GRCm38
          Downstream FGGY carbohydrate kinase domain containing Fggy 2310009E04Rik 88.2kb Anti-sense Chr. 4 - 95557485 - 95926939 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 562958 AK086876.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 192341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170129 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106991 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03880894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03594091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03540647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03413685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03668231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03716247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03432741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03641191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03432733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03769038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03413688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03766011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03432744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03861872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03423768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23123014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23122970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03423770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03969140 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0