Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU022754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040433.1 | 1 - 2 | 273 | 68 | ||
GenBank mRNA | AK159003.1 | - | 1 - 2 | 274 | 68 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
wingless-type MMTV integration site family, member 7B |
Wnt7b | Wnt-7b | Anti-sense | Chr. 15 - 85535437 - 85581821 on NCBI Build GRCm38 | |
Downstream | long intergenic noncoding RNA near Ppara | Lincppara |
Mirlet7bhg, linc-Ppara |
119.3kb | Sense | Chr. 15 - 85703773 - 85707524 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |