Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03951275_m1
          See other GM20756 GE Assays ›
          Gene Symbol:
          Gm20756
          Gene Name:
          predicted gene, 20756
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 25931426 - 25977227 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03951275_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm20756
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040771.1 8 - 9 849 96
          GenBank mRNA AK165854.1 - 8 - 9 850 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protection of telomeres 1A Pot1a 1500031H18Rik,
          AI851169,
          Pot1
          -163.4kb Anti-sense Chr. 6 - 25743735 - 25809277 on NCBI Build GRCm38
          Downstream glutamate receptor,
          metabotropic 8
          Grm8 A230002O04,
          Gprc1h,
          mGluR8
          1.3mb Anti-sense Chr. 6 - 27275121 - 28135837 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000210.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 194828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848285 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 7027 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191378 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05539888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05043614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05687770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05667943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05337996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05862005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05758673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05043615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05734627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05170501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05844811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05862006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05734616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24308675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05337997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600001328.mm.1 TC0600000210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001328.mm.1 NR_040771.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001328.mm.1 NR_040771.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001331.mm.1 TC0600000210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001331.mm.1 NR_040771.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001331.mm.1 NR_040771.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001334.mm.1 TC0600000210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001334.mm.1 NR_040771.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001334.mm.1 NR_040771.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001333.mm.1 TC0600000210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001333.mm.1 NR_040771.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001333.mm.1 NR_040771.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001329.mm.1 TC0600000210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001329.mm.1 NR_040771.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001329.mm.1 NR_040771.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0