Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03951321_m1
          See other AA536875 GE Assays ›
          Gene Symbol:
          AA536875
          Gene Name:
          expressed sequence AA536875
          Gene Aliases:
          Gm1591
          Chromosome Location:
          Chr.14: 123168503 - 123176176 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03951321_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA536875
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045144.1 2 - 3 327 91
          NR_045145.1 3 - 4 417 91
          GenBank mRNA AK166554.1 - 2 - 3 333 91
          CF172390.1 - 3 - 4 417 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY735584.1 - 2 - 3 333 Chr. 14 - 123169182 - 123174970 [+] on Build GRCm38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream transmembrane and tetratricopeptide repeat containing 4 Tmtc4 4930403J22Rik,
          5730419O14Rik
          -186.4kb Anti-sense Chr. 14 - 122918971 - 122983283 on NCBI Build GRCm38
          Downstream sodium leak channel,
          non-selective
          Nalcn A530023G15Rik,
          AI849508,
          Vgcnl1
          102.0kb Anti-sense Chr. 14 - 123276634 - 123627363 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001345.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC1500001216.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11719738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11830062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11464758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11170495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11411187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11142143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29965603 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400010064.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010064.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010064.mm.1 NR_045145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010067.mm.1 NR_045145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010070.mm.1 NR_045145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010072.mm.1 NR_045145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010066.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010066.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010066.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010066.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010066.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010071.mm.1 NR_045145.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 TC1400001345.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045143.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045143.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045144.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045144.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045145.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400010068.mm.1 NR_045145.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0