Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952085_m1
          See other GM6567 GE Assays ›
          Gene Symbol:
          Gm6567
          Gene Name:
          predicted gene 6567
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 73097752 - 73102068 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952085_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm6567
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046024.1 1 - 2 1418 71
          GenBank mRNA BC052231.1 - 1 - 2 1418 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream multiple C2 domains,
          transmembrane 2
          Mctp2 Gm489 -792.6kb Sense Chr. 7 - 72077827 - 72306595 on NCBI Build GRCm38
          Downstream predicted gene,
          20083
          Gm20083 81.5kb Anti-sense Chr. 7 - 73183605 - 73192432 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003489.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P129643 BC052231.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 2604 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P129643 BC052231.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129643 BC052231.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06036325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05996122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05895816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05947125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05996117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06036330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05916485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25227112 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700029598.mm.1 TC0700003489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029598.mm.1 NR_046024.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029598.mm.1 NR_046024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029599.mm.1 TC0700003489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029599.mm.1 NR_046024.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029599.mm.1 NR_046024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029593.mm.1 TC0700003489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029593.mm.1 NR_046024.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029593.mm.1 NR_046024.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029595.mm.1 TC0700003489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029595.mm.1 NR_046024.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029595.mm.1 NR_046024.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 571793 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-777976674b-tw4cl:80/100.66.134.228:80.
          git-commit: 6354fc3e62d97773410f28d6123c879be4bd0e1c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0