Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03952990_s1
          See other 2610105M22RIK GE Assays ›
          Gene Symbol:
          2610105M22Rik
          Gene Name:
          RIKEN cDNA 2610105M22 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03952990_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610105M22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK011823.1 - 1 1052 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 4933422A05 gene 4933422A05Rik -76.5kb Anti-sense Chr. 9 - 35281105 - 35285423 on NCBI Build GRCm38
          Downstream cell adhesion molecule-related/down-regulated by oncogenes Cdon CDO,
          Cd0,
          Orcam
          58.0kb Anti-sense Chr. 9 - 35421129 - 35507652 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07813112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07813540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26422250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432773_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1432774_PM_at - -
          Mouse Genome 430 2.0 Array 1432774_PM_at - -
          Mouse Genome 430 2.0 Array 1432773_PM_at - -
          Mouse Genome 430 2.0 Array 1432774_at - -
          Mouse Genome 430 2.0 Array 1432773_at - -
          Mouse Expression Set 430 1432774_PM_at - -
          Mouse Expression Set 430 1432773_at - -
          Mouse Expression Set 430 1432774_at - -
          Mouse Expression Set 430 1432773_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432774_at - -
          Affymetrix HT Mouse Genome 430B Array 1432773_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432773_at - -
          Affymetrix HT Mouse Genome 430B Array 1432774_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0