Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03953319_s1
          See other 2810001A02RIK GE Assays ›
          Gene Symbol:
          2810001A02Rik
          Gene Name:
          RIKEN cDNA 2810001A02 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03953319_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2810001A02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK012625.1 - 451 67
          BE952576.1 - 67
          BG074436.2 - 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BX631349.1 - 1 Chr. 15 - 86213775 - 86214044 [+] on Build GRCm38 67
          GenBankmRNA CX234730.1 - 1 443 Chr. 15 - 86213786 - 86214360 [-] on Build GRCm38 67
          GenBankmRNA CK793247.1 - 1 436 Chr. 15 - 86213754 - 86214341 [-] on Build GRCm38 67
          GenBankmRNA BP776142.1 - 1 Chr. 15 - 86213754 - 86214314 [+] on Build GRCm38 67
          GenBankmRNA BY585924.1 - 1 241 Chr. 15 - 86213756 - 86214158 [-] on Build GRCm38 67
          GenBankmRNA W41038.1 - 434 Chr. - - [-] on Build 67
          GenBankmRNA AA434975.1 - 1 411 Chr. 15 - 86213889 - 86214329 [-] on Build GRCm38 67
          GenBankmRNA EG564690.1 - Chr. - - [-] on Build 67
          GenBankmRNA BM937289.1 - 1 364 Chr. 15 - 86213617 - 86214281 [-] on Build GRCm38 67
          GenBankmRNA CK032080.1 - 1 437 Chr. 15 - 86213630 - 86214354 [-] on Build GRCm38 67
          GenBankmRNA BY606671.1 - 1 225 Chr. 15 - 86213774 - 86214142 [-] on Build GRCm38 67
          GenBankmRNA BX515995.1 - 1 320 Chr. 15 - 86213817 - 86214237 [-] on Build GRCm38 67
          GenBankmRNA BF585342.1 - 1 468 Chr. 15 - 86213827 - 86214385 [-] on Build GRCm38 67
          GenBankmRNA W82199.1 - 452 Chr. - - [-] on Build 67
          GenBankmRNA AW553328.2 - 1 Chr. 15 - 86213776 - 86214168 [+] on Build GRCm38 67
          GenBankmRNA BY584431.1 - 1 181 Chr. 15 - 86213762 - 86214098 [-] on Build GRCm38 67
          GenBankmRNA AA792477.1 - 1 110 Chr. 15 - 86213548 - 86214012 [-] on Build GRCm38 67
          GenBankmRNA AW989427.1 - 1 389 Chr. 15 - 86213815 - 86214306 [-] on Build GRCm38 67
          GenBankmRNA BE946816.1 - 1 Chr. 15 - 86213736 - 86214139 [+] on Build GRCm38 67
          GenBankmRNA BE634544.1 - 1 Chr. 15 - 86213776 - 86214203 [+] on Build GRCm38 67
          GenBankmRNA AI426276.1 - 1 Chr. 15 - 86213773 - 86214079 [+] on Build GRCm38 67
          GenBankmRNA EG563941.1 - 1 Chr. 15 - 86213773 - 86214133 [+] on Build GRCm38 67
          GenBankmRNA BG087067.2 - 1 446 Chr. 15 - 86213773 - 86214363 [-] on Build GRCm38 67
          GenBankmRNA AI646166.1 - 1 Chr. 15 - 86213775 - 86214079 [+] on Build GRCm38 67
          GenBankmRNA BG971627.1 - 1 445 Chr. 15 - 86213773 - 86214363 [-] on Build GRCm38 67
          GenBankmRNA AA097377.1 - 1 377 Chr. 15 - 86213859 - 86214295 [-] on Build GRCm38 67
          GenBankmRNA BE133921.1 - 1 Chr. 15 - 86213775 - 86214267 [+] on Build GRCm38 67
          GenBankmRNA BF539835.1 - 1 430 Chr. 15 - 86213773 - 86214342 [-] on Build GRCm38 67
          GenBankmRNA CN539193.1 - 1 426 Chr. 15 - 86213534 - 86214343 [-] on Build GRCm38 67
          GenBankmRNA AA763674.1 - 1 413 Chr. 15 - 86213843 - 86214333 [-] on Build GRCm38 67
          GenBankmRNA BE635053.1 - 1 378 Chr. 15 - 86213823 - 86214295 [-] on Build GRCm38 67
          GenBankmRNA BQ892173.1 - 1 442 Chr. 15 - 86213421 - 86214359 [-] on Build GRCm38 67
          GenBankmRNA AI482522.1 - 1 Chr. 15 - 86213773 - 86214203 [+] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ceramide kinase Cerk AI848610,
          D330016D08Rik,
          mKIAA1646
          -27.6kb Sense Chr. 15 - 86139101 - 86186193 on NCBI Build GRCm38
          Downstream TBC1 domain family,
          member 22a
          Tbc1d22a BC023106,
          D15Ertd781e
          58.0 Anti-sense Chr. 15 - 86214421 - 86498503 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103269 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431283 - -
          Affymetrix HT MG-430 PM Array Plate 1436323_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436322_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431282 - -
          Mouse Genome 430 2.0 Array 1436322_a_at - -
          Mouse Genome 430 2.0 Array 1436323_at - -
          Mouse Genome 430 2.0 Array 1436322_PM_a_at - -
          Mouse Genome 430 2.0 Array 1436323_PM_at - -
          Mouse Expression Set 430 1436322_PM_a_at - -
          Mouse Expression Set 430 1436323_at - -
          Mouse Expression Set 430 1436322_a_at - -
          Mouse Expression Set 430 1436323_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436323_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436323_at - -
          Affymetrix HT Mouse Genome 430B Array 1436322_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1436322_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 741113 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100610398 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix