Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03954556_s1
          See other 3110021N24RIK GE Assays ›
          Gene Symbol:
          3110021N24Rik
          Gene Name:
          RIKEN cDNA 3110021N24 gene
          Gene Aliases:
          2610002D03Rik
          Chromosome Location:
          Chr.4: 108719649 - 108781904 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03954556_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 3110021N24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001254730.1 NP_001241659.1 4 1013 77
          GenBank mRNA AK014069.1 - 1 1047 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF471482.1 - 1 195 Chr. 4 - 108780911 - 108781409 [+] on Build GRCm38 77
          GenBankmRNA BB873185.1 - 1 159 Chr. 4 - 108780948 - 108781290 [+] on Build GRCm38 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger,
          FYVE domain containing 9
          Zfyve9 E030027L17,
          Madhip,
          NSP,
          SARA,
          SMADIP
          Anti-sense Chr. 4 - 108637834 - 108781148 on NCBI Build GRCm38
          Downstream basic transcription factor 3-like 4 Btf3l4 4632412E09Rik,
          5730434I03Rik,
          AI316863,
          AU022207,
          AW552446
          33.0kb Anti-sense Chr. 4 - 108814295 - 108834297 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001153.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137319 - -
          Whole Mouse Genome G4122A A_52_P75220 AK011275.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03404460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03380074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03387657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03555862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03235028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03875641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03235025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03235024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03931539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03848669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03890327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03235038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197655 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453764_PM_at - -
          Mouse Genome 430 2.0 Array 1453764_PM_at - -
          Mouse Genome 430 2.0 Array 1453764_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73133_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73133_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400008263.mm.1 TC0400001153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008261.mm.1 TC0400001153.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008262.mm.1 TC0400001153.mm.1 MTA TC
          Mouse Expression Set 430 1453764_PM_at - -
          Mouse Expression Set 430 1453764_at - -
          Affymetrix HT Mouse Genome 430B Array 1453764_at - -
          Affymetrix HT Mouse Genome 430B Array 1453764_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106350563 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259861 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0