Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03955567_m1
          See other 4921511C10RIK GE Assays ›
          Gene Symbol:
          4921511C10Rik
          Gene Name:
          RIKEN cDNA 4921511C10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 79212919 - 79253913 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03955567_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4921511C10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045502.1 2 - 3 1026 90
          GenBank mRNA AK014864.1 - 2 - 3 1026 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glyceraldehyde-3-phosphate dehydrogenase pseudogene Gm10291 ENSMUSG00000070443 -329.7kb Sense Chr. 3 - 78917240 - 78918459 on NCBI Build GRCm38
          Downstream predicted gene,
          17359
          Gm17359 91.4kb Sense Chr. 3 - 79345321 - 79464129 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000619.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2341890 AK137931.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 208940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 446250 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 551898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 54935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28839 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5720 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04959437 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04959384 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02842904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02395278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02332862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02433544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02538294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02842910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02393978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02842916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02433526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22262347 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432070_PM_at - -
          Mouse Genome 430 2.0 Array 1432070_at - -
          Mouse Genome 430 2.0 Array 1432070_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300004055.mm.1 TC0300000619.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004055.mm.1 NR_045502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004055.mm.1 NR_045502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004057.mm.1 TC0300000619.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004057.mm.1 NR_045502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004057.mm.1 NR_045502.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004056.mm.1 TC0300000619.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300004056.mm.1 NR_045502.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300004056.mm.1 NR_045502.1 Refseq
          Mouse Expression Set 430 1432070_PM_at - -
          Mouse Expression Set 430 1432070_at - -
          Affymetrix HT Mouse Genome 430B Array 1432070_at - -
          Affymetrix HT Mouse Genome 430B Array 1432070_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 335732 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102100519 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0