Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956493_m1
          See other A030001D20RIK GE Assays ›
          Gene Symbol:
          A030001D20Rik
          Gene Name:
          RIKEN cDNA A030001D20 gene
          Gene Aliases:
          1110025D03Rik
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956493_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A030001D20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003910.1 - 1 - 2 59 105
          AK037147.1 - 1 - 2 75 105
          AK049852.1 - 1 - 2 61 105
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB630270.1 - 1 - 2 75 Chr. 7 - 45636556 - 45638758 [-] on Build GRCm38 105
          GenBankmRNA CK031458.1 - 1 - 2 59 Chr. 7 - 45636472 - 45638736 [-] on Build GRCm38 105
          GenBankmRNA BB655923.1 - 1 - 2 72 Chr. 7 - 45636558 - 45638755 [-] on Build GRCm38 105
          GenBankmRNA CX202519.1 - 1 - 2 66 Chr. 7 - 45636407 - 45638750 [-] on Build GRCm38 105
          GenBankmRNA AA734726.1 - 1 - 2 61 Chr. 7 - 45636793 - 45638744 [-] on Build GRCm38 105
          GenBankmRNA CB589422.1 - 1 - 2 81 Chr. 7 - 45636424 - 45638765 [-] on Build GRCm38 105
          GenBankmRNA BU561856.1 - 68 Chr. - - [-] on Build 105
          GenBankmRNA BY704165.1 - 1 - 2 59 Chr. 7 - 45636217 - 45638742 [-] on Build GRCm38 105
          GenBankmRNA BB597893.2 - 1 - 2 61 Chr. 7 - 45636547 - 45638744 [-] on Build GRCm38 105

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream izumo sperm-egg fusion 1 Izumo1 1700058F15Rik,
          Izumo
          -8.9kb Anti-sense Chr. 7 - 45621811 - 45627242 on NCBI Build GRCm38
          Downstream MEF2 activating motif and SAP domain containing transcriptional regulator Mamstr 2810022D01Rik,
          5430432N15Rik,
          AW743872,
          Mastr
          458.0 Anti-sense Chr. 7 - 45639200 - 45648206 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2213214 AK037147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2213214 AK037147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209578 AK037147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P209578 AK003910.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1429258_PM_at - -
          Mouse Genome 430 2.0 Array 1429258_at - -
          Mouse Genome 430 2.0 Array 1429258_PM_at - -
          Mouse Expression Set 430 1429258_PM_at - -
          Mouse Expression Set 430 1429258_at - -
          Affymetrix HT Mouse Genome 430B Array 1429258_at - -
          Affymetrix HT Mouse Genome 430B Array 1429258_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 315496 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0