Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956541_s1
          See other ITPKB GE Assays ›
          Gene Symbol:
          Itpkb
          Gene Name:
          inositol 1,4,5-trisphosphate 3-kinase B
          Gene Aliases:
          1110033J02Rik, E130307H12Rik
          Chromosome Location:
          Chr.1: 180330476 - 180424802 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956541_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itpkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK004067.1 - 1 149 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CJ260572.1 - 1 306 Chr. 1 - 180424355 - 180424801 [+] on Build GRCm38 88
          GenBankmRNA CJ260783.1 - 1 333 Chr. 1 - 180424328 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA BQ175413.1 - 1 Chr. 1 - 180424543 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA W82678.1 - 178 Chr. - - [-] on Build 88
          GenBankmRNA BM053199.1 - 1 Chr. 1 - 180424506 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA CJ259332.1 - 1 323 Chr. 1 - 180424338 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA BG793645.1 - 1 Chr. 1 - 180424431 - 180424802 [-] on Build GRCm38 89
          GenBankmRNA CJ260992.1 - 1 297 Chr. 1 - 180424364 - 180424801 [+] on Build GRCm38 88
          GenBankmRNA AA939529.1 - 1 210 Chr. 1 - 180424451 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA CJ259474.1 - 1 312 Chr. 1 - 180424349 - 180424801 [+] on Build GRCm38 88
          GenBankmRNA BE196957.1 - 1 Chr. 1 - 180424322 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA BY704314.1 - 1 149 Chr. 1 - 180424512 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA BQ174972.1 - 1 Chr. 1 - 180424543 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA BI790334.1 - 1 Chr. 1 - 180424359 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA CJ259631.1 - 1 317 Chr. 1 - 180424344 - 180424801 [+] on Build GRCm38 88
          GenBankmRNA AA170321.1 - 1 129 Chr. 1 - 180424532 - 180424802 [+] on Build GRCm38 88
          GenBankmRNA AW048732.1 - 1 Chr. 1 - 180424408 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA AW121826.1 - 1 Chr. 1 - 180424409 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA BE951232.1 - 1 Chr. 1 - 180424428 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA CJ259461.1 - 1 281 Chr. 1 - 180424380 - 180424802 [+] on Build GRCm38 88
          GenBankmRNA BX638361.1 - 1 Chr. 1 - 180424550 - 180424802 [-] on Build GRCm38 88
          GenBankmRNA CF583201.1 - Chr. - - [-] on Build 88
          GenBankmRNA CJ260534.1 - 1 277 Chr. 1 - 180424384 - 180424802 [+] on Build GRCm38 88
          GenBankmRNA CJ259441.1 - 1 324 Chr. 1 - 180424337 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA BI854404.1 - 1 507 Chr. 1 - 180424153 - 180424802 [+] on Build GRCm38 88
          GenBankmRNA CJ258648.1 - 1 344 Chr. 1 - 180424317 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA CJ260363.1 - 1 311 Chr. 1 - 180424350 - 180424798 [+] on Build GRCm38 88
          GenBankmRNA CJ261029.1 - 1 302 Chr. 1 - 180424359 - 180424801 [+] on Build GRCm38 88
          GenBankmRNA CJ258917.1 - 1 326 Chr. 1 - 180424335 - 180424801 [+] on Build GRCm38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream glyceraldehyde-3-phosphate dehydrogenase pseudogene Gm5069 EG277333 -94.0kb Anti-sense Chr. 1 - 180326969 - 180330549 on NCBI Build GRCm38
          Downstream RIKEN cDNA 6330403A02 gene 6330403A02Rik ENSMUSG00000056615,
          Gm10001,
          R75555
          7.6kb Anti-sense Chr. 1 - 180432373 - 180483519 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          ABI

          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 411745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0