Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956918_s1
          See other 1700039O17RIK GE Assays ›
          Gene Symbol:
          1700039O17Rik
          Gene Name:
          RIKEN cDNA 1700039O17 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956918_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700039O17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK007166.1 - 1 256 93
          AK018843.1 - 1 250 93
          AK018940.1 - 1 251 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CN836158.1 - 1 291 Chr. 2 - 173291220 - 173291605 [+] on Build GRCm38 93
          GenBankmRNA AV209124.2 - Chr. - - [-] on Build 93
          GenBankmRNA BY707206.1 - 1 246 Chr. 2 - 173291263 - 173291605 [+] on Build GRCm38 93
          GenBankmRNA CR520047.1 - 1 Chr. 2 - 173291286 - 173291605 [-] on Build GRCm38 93
          GenBankmRNA BY707250.1 - 1 255 Chr. 2 - 173291256 - 173291570 [+] on Build GRCm38 93
          GenBankmRNA BY706636.1 - 1 246 Chr. 2 - 173291263 - 173291605 [+] on Build GRCm38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream prostate transmembrane protein,
          androgen induced 1
          Pmepa1 2210418I02Rik,
          AW455466,
          Erg1.2,
          N4wbp4,
          Stag1,
          Tmepai
          -14.7kb Anti-sense Chr. 2 - 173224458 - 173276533 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700021F07 gene 1700021F07Rik 231.0kb Sense Chr. 2 - 173522592 - 173528502 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Whole Mouse Genome G4122A A_51_P413725 AK018843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413725 AK018940.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01600249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01786714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02071206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02127897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02154290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01696327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01600244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01600246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21868893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02127894 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102120605 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-54d969bb78-8h2jf:80/100.66.130.62:80.
          git-commit: 3e69e100148d4b3be95fd74bb38be07b481340d5
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0