Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956943_s1
          See other 1700084E18RIK GE Assays ›
          Gene Symbol:
          1700084E18Rik
          Gene Name:
          RIKEN cDNA 1700084E18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 30237197 - 30237631 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956943_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700084E18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028299.1 1 258 68
          GenBank mRNA AK006990.1 - 1 259 68
          AK018989.1 - 1 147 68
          AK018997.1 - 1 147 68
          AK034708.1 - 852 68
          BC062268.1 - 1 234 68
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707421.1 - 1 147 Chr. 2 - 30237191 - 30237519 [-] on Build GRCm38 68
          GenBankmRNA BY707385.1 - 1 147 Chr. 2 - 30237191 - 30237519 [-] on Build GRCm38 68
          GenBankmRNA BE197082.1 - 1 Chr. 2 - 30237200 - 30237561 [+] on Build GRCm38 68
          GenBankmRNA BX636333.1 - 1 Chr. 2 - 30237197 - 30237475 [+] on Build GRCm38 68
          GenBankmRNA BB090636.2 - 1 81 Chr. 2 - 30236799 - 30237454 [-] on Build GRCm38 68
          GenBankmRNA BY707046.1 - 1 259 Chr. 2 - 30237200 - 30237631 [-] on Build GRCm38 68
          GenBankmRNA BE137757.1 - 1 234 Chr. 2 - 30237215 - 30237607 [-] on Build GRCm38 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cysteine conjugate-beta lyase 1 Ccbl1 2010009K05Rik,
          AI182306,
          Gtk,
          Kat1,
          KatI
          -30.9kb Sense Chr. 2 - 30185124 - 30206295 on NCBI Build GRCm38
          Downstream leucine rich repeat containing 8A Lrrc8a Lrrc8,
          mKIAA1437
          250.0 Anti-sense Chr. 2 - 30237769 - 30263790 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003266.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P370746 AK034708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P370746 AK034708.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103025 - -
          Whole Mouse Genome G4122A A_52_P370746 AK034708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P370746 AK034708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P520583 NR_028299.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P520583 AK006990.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P520583 AK018989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P520583 AK018997.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P520583 AK034708.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100459 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481452 - -
          Affymetrix Mouse Gene 1.1 ST Array 10481451 - -
          Affymetrix Mouse Clariom S Array PSR0200027607.mm.1 TC0200003266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027607.mm.1 NR_028299.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027607.mm.1 NR_028299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027608.mm.1 TC0200003266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027608.mm.1 NR_028299.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027608.mm.1 NR_028299.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027606.mm.1 TC0200003266.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027606.mm.1 NR_028299.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027606.mm.1 NR_028299.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436594 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix