Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03956994_g1
          See other 2700046G09RIK GE Assays ›
          Gene Symbol:
          2700046G09Rik
          Gene Name:
          RIKEN cDNA 2700046G09 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 32389216 - 32391184 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03956994_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2700046G09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033198.1 1 - 2 166 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY711528.1 - 1 - 2 158 Chr. 19 - 32389224 - 32390134 [+] on Build GRCm38 93
          GenBankmRNA BY711737.1 - 1 - 2 158 Chr. 19 - 32389224 - 32390138 [+] on Build GRCm38 93
          GenBankmRNA BB628316.1 - 1 - 2 165 Chr. 19 - 32389217 - 32390168 [+] on Build GRCm38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream N-acylsphingosine amidohydrolase 2 Asah2 AI585898 -281.0kb Anti-sense Chr. 19 - 31982597 - 32108233 on NCBI Build GRCm38
          Downstream multiple inositol polyphosphate histidine phosphatase 1 Minpp1 AA408516 95.6kb Sense Chr. 19 - 32485769 - 32515370 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000464.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of myelination
          RNA stabilization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030010 NR_033198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2030010 AK035700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110875 NR_033198.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110875 AK012385.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110875 AK019229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2110875 AK035700.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144347 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144347 - -
          Whole Mouse Genome G4122A A_52_P324656 NR_033198.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324656 AK012385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324656 AK019229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324656 AK035700.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324656 AK170065.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114666 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14375665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14375664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32070045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431163_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10462501 - -
          Mouse Genome 430 2.0 Array 1431163_at - -
          Mouse Genome 430 2.0 Array 1431163_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67188_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67188_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900004238.mm.1 TC1900000464.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004238.mm.1 NR_033198.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004238.mm.1 NR_033198.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004239.mm.1 TC1900000464.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004239.mm.1 NR_033198.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004239.mm.1 NR_033198.1 Refseq
          Mouse Expression Set 430 1431163_PM_at - -
          Mouse Expression Set 430 1431163_at - -
          Affymetrix HT Mouse Genome 430B Array 1431163_at - -
          Affymetrix HT Mouse Genome 430B Array 1431163_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 339417 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104150022 - -
          Illumina mouse-6 v1.0 expression beadchip 104150022 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2546887 - -
          Illumina Sentrix mouse-6.v1 bead array 104150022 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104150022 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2546887 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0