Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03957322_g1
          See other 4930538L07RIK GE Assays ›
          Gene Symbol:
          4930538L07Rik
          Gene Name:
          RIKEN cDNA 4930538L07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 47757041 - 47757519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03957322_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930538L07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019723.1 - 1 - 2 194 64
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY715567.1 - 1 - 2 194 Chr. 14 - 47757034 - 47757498 [-] on Build GRCm38 64
          GenBankmRNA CR521426.1 - 1 - 2 Chr. 14 - 47757044 - 47757436 [+] on Build GRCm38 64

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream kinectin 1 Ktn1 -20.5kb Anti-sense Chr. 14 - 47648453 - 47736564 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930447J18 gene 4930447J18Rik 4930425M13Rik 141.4kb Anti-sense Chr. 14 - 47898864 - 47944189 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11771158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11444275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11128390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11798744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11473022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11587539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11128397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583999 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11254380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11473020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11771160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29584001 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11254379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29583949 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 441399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0