Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9230118N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020358.1 | - | 1 | 1150 | 85 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 1962 | Mir1962 |
mir-1962, mmu-mir-1962 |
-575.0 | Sense | Chr. 7 - 135566162 - 135566282 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 5830432E09 gene | 5830432E09Rik | 80.3kb | Anti-sense | Chr. 7 - 135648457 - 135652314 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |