Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03958378_s1
          See other 9430007M09RIK GE Assays ›
          Gene Symbol:
          9430007M09Rik
          Gene Name:
          RIKEN cDNA 9430007M09 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03958378_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9430007M09Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020407.1 - 1 651 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream frizzled homolog 9 (Drosophila) Fzd9 mfz9 -13.3kb Anti-sense Chr. 5 - 135248938 - 135251047 on NCBI Build GRCm38
          Downstream FK506 binding protein 6 Fkbp6 1700008G22Rik,
          36kDa,
          AU017274,
          D5Ertd724e,
          FKBP-36,
          FKBP-6
          26.6kb Anti-sense Chr. 5 - 135291704 - 135351807 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241134 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04435379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04533689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04498265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04533694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04591579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04880203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04533690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04533695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04137992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04216255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04247795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24115587 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline