Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03979560_m1
          See other GM19757 GE Assays ›
          Gene Symbol:
          Gm19757
          Gene Name:
          predicted gene, 19757
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 105481266 - 105492668 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03979560_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm19757
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040297.1 2 - 3 464 76
          GenBank mRNA AK145340.1 - 2 - 3 465 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CO795598.1 - 465 Chr. - - [-] on Build 76
          GenBankmRNA BY735554.1 - 2 - 3 462 Chr. 6 - 105483506 - 105492665 [-] on Build GRCm38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream contactin 6 Cntn6 NB-3 -617.9kb Anti-sense Chr. 6 - 104492704 - 104863406 on NCBI Build GRCm38
          Downstream predicted gene 15631 Gm15631 OTTMUSG00000024600 186.1kb Sense Chr. 6 - 105671197 - 105679405 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002875.mm.1
          This assay targets the same location as RTA transcript cluster(s) TC0400003485.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434863 AK002426.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434863 AK013210.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434863 AK132102.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434863 BC024341.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434863 NM_025797.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06013453 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06013453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05800762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05168740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05571879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05874219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825495 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05497024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05874212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05297643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24718022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05254027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24717987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05800772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600022070.mm.1 TC0600002875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022070.mm.1 NR_040297.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022070.mm.1 NR_040297.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022068.mm.1 TC0600002875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022068.mm.1 NR_040297.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022068.mm.1 NR_040297.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022069.mm.1 TC0600002875.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022069.mm.1 NR_040297.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022069.mm.1 NR_040297.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 341143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0