Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03992852_s1
          See other CLCA4B GE Assays ›
          Gene Symbol:
          Clca4b
          Gene Name:
          chloride channel calcium activated 4B
          Gene Aliases:
          AI747448, Clca7
          Chromosome Location:
          Chr.3: 144910921 - 144932529 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03992852_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033199.3 NP_001028371.1 14 2846 153
          GenBank mRNA AK136515.1 - 14 2851 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003082.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          chloride transmembrane transport
          molecular_function
          intracellular calcium activated chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 BC139088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 BC139089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 NM_001033199.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 BC139088.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 BC139089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 NM_001033199.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152097 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123432 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 BC139088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 BC139089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 NM_001033199.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 AK136515.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 BC139088.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 BC139089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 NM_001033199.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 BC139088.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 BC139089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 NM_001033199.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 BC139088.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 BC139089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 NM_001033199.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P95161 AK136515.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105426 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105408 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123432 - -
          Whole Mouse Genome G4122A A_51_P134993 AK054018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134993 AK033591.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P134993 NM_207208.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02301351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03086210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02857939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605685 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502620 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502621 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502615 - -
          Affymetrix HT MG-430 PM Array Plate 1439796_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502606 - -
          Affymetrix Mouse Gene 1.1 ST Array 10502613 - -
          Mouse Genome 430 2.0 Array 1439796_at - -
          Mouse Genome 430 2.0 Array 1439796_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99709_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 99709_at - -
          Affymetrix Mouse Clariom S Array PSR0300024235.mm.1 TC0300003082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024235.mm.1 NM_001033199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024235.mm.1 NM_001033199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024233.mm.1 TC0300003082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024233.mm.1 NM_001033199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024233.mm.1 NM_001033199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024234.mm.1 TC0300003082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024234.mm.1 NM_001033199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024234.mm.1 NM_001033199.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024236.mm.1 TC0300003082.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024236.mm.1 NM_001033199.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024236.mm.1 NM_001033199.3 Refseq
          Mouse Expression Set 430 1447394_PM_at - -
          Mouse Expression Set 430 1439796_PM_at - -
          Mouse Expression Set 430 1439796_at - -
          Mouse Expression Set 430 1447394_at - -
          Affymetrix HT Mouse Genome 430B Array 1439796_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439796_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 577451 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253045 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2844353 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1253045 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2844353 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0