Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900008261.mm.1 |
TC0900001049.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900008261.mm.1 |
NM_001243092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900008261.mm.1 |
NM_001243092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022441.mm.1 |
TC0900002719.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022441.mm.1 |
NM_008182.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022441.mm.1 |
NM_008182.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900008274.mm.1 |
TC0900001050.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900008274.mm.1 |
NM_001122660.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900008274.mm.1 |
NM_001122660.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900008250.mm.1 |
TC0900001049.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900008250.mm.1 |
NM_008181.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900008250.mm.1 |
NM_008181.3 |
Refseq |